| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037292.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037292.hg.1 |
ENST00000545382.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037292.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037292.hg.1 |
TC03002108.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300200872.hg.1 |
TC0300013520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000295522.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
ENST00000490800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013520.hg.1 |
NM_021101.4 |
Refseq |