Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00331221_s1
          See other LOC151657 GE Assays ›
          Gene Symbol:
          LOC151657
          Gene Name:
          uncharacterized LOC151657
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00331221_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC151657
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK057598.1 - 1 1453 93
          AK091114.1 - 1 530 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 168636 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 353298 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 52466 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 94868 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 15842 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3436646 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 36941 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3436646 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P16351398 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16351398 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16351398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560928_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300011935.hg.1 ENST00000624541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011935.hg.1 ENST00000624541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011935.hg.1 ENST00000624541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011935.hg.1 ENST00000624541.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1560928_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560928_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560928_at1 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline