Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00383084_s1
          See other LINC01521 GE Assays ›
          Gene Symbol:
          LINC01521
          Gene Name:
          long intergenic non-protein coding RNA 1521
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 31346759 - 31351155 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00383084_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01521
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_120386.1 1 478 118
          GenBank mRNA AK000471.1 - 1 348 118
          CR456348.1 - 1 351 118

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK000471.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32155735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32155737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32155738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11473898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32155739 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 29478 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 30338 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 576289 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P57480 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P57482 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5197 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 42947 - -
          Agilent Human Genome CGH Microarray 44B A_14_P116407 - -
          Agilent Human Genome CGH Microarray 44A A_14_P116407 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37483 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3298455 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P57482 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P57480 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P116407 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P116407 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P57480 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P57482 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312364.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312365.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312366.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312367.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312368.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312369.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312370.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312371.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312372.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY312373.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY321515.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 AY321516.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 BC031961.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 BC130477.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112680 U85653.1 genbank_mrna_human
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 58701 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P116407 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P116407 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8072487 - -
          Affymetrix Human Gene 1.1 ST Array 8072486 - -
          Affymetrix HT HG-U133+ PM Array Plate 224073_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 ENST00000614284.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149862.hg.1 TC2200007101.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200149862.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149862.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149862.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149862.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 ENST00000614284.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007101.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149864.hg.1 TC2200007101.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200149864.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149864.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149864.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149864.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 TC2200007101.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 ENST00000614284.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 NR_120386.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 ENST00000504184.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 ENST00000614284.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149863.hg.1 NR_120386.1 Refseq
          Human Genome U133 Plus 2.0 Array 224073_at - -
          Human Genome U133 Plus 2.0 Array 224073_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224073_at8 - -
          Human Genome U95 Set 77107_at - -
          Human Genome U95 Set 77107_PM_at - -
          Human Genome U133 Set 224073_PM_at - -
          Human Genome U133 Set 224073_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 167741 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1702757 BC004550.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1702757 AK000471.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1702757 BC004550.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1702757 AK000471.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1702757 BC004550.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1702757 AK000471.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1702757 BC004550.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1702757 AK000471.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1702757 BC004550.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1702757 AK000471.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline