Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00395151_m1
          See other ADARB2-AS1 GE Assays ›
          Gene Symbol:
          ADARB2-AS1
          Gene Name:
          ADARB2 antisense RNA 1
          Gene Aliases:
          C10orf109, NCRNA00168, bA466B20.1
          Chromosome Location:
          Chr.10: 1526630 - 1556984 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00395151_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADARB2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033387.1 1 - 2 264 63
          GenBank mRNA AF086274.1 - 1 - 2 264 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000006484.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000013.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P29954 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P29954 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33194 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13735 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P29954 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P29954 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P29954 - -
          Agilent Human 2 cDNA Microarray (G4101A) 842573 - -
          Agilent Human 2 cDNA Microarray (G4101A) 842573 - -
          Agilent Human 1B Microarray G4111A A_32_P29963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561127_PM_at - -
          Affymetrix Human Genome U219 Array 11759262_PM_at - -
          Affymetrix Human Genome U219 Array 11759262_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR10000161.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10000161.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10000161.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10000161.hg.1 TC10000013.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000006484.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10000160.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10000160.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10000160.hg.1 TC10000013.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10000164.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10000164.hg.1 NR_033387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10000164.hg.1 TC10000013.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000144462.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144462.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144462.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144462.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144462.hg.1 TC1000006484.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144457.hg.1 TC1000006484.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000144456.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144456.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144456.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000144456.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000144456.hg.1 TC1000006484.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000381301.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 ENST00000421697.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 NR_033387.1 Refseq
          Affymetrix Human Clariom D Assay TC1000006484.hg.1 NR_033387.1 Refseq
          Human Genome U133 Plus 2.0 Array 1561127_at - -
          Human Genome U133 Plus 2.0 Array 1561127_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561127_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043510 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043510 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 184737 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1712854 NR_033387.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1712854 AF086274.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1712854 NR_033387.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1712854 AF086274.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1712854 NR_033387.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1712854 AF086274.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline