Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00411285_g1
          See other LINC00307 GE Assays ›
          Gene Symbol:
          LINC00307
          Gene Name:
          long intergenic non-protein coding RNA 307
          Gene Aliases:
          D21S2091E, NCRNA00307
          Chromosome Location:
          Chr.21: 30209151 - 30211783 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00411285_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00307
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038855.1 2 - 3 240 74
          GenBank mRNA AY063455.1 - 2 - 3 240 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007880.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000362.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11344174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11344168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32003663 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 73940 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 259710 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 114853 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 219322 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 74733 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 191922 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 44296 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 298897 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 169841 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 330816 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3492042 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32804 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18593 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3492042 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3492042 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P212624 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41428068 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21231298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563385_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 NR_038855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 NR_038855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007880.hg.1 NR_038855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21004138.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21004138.hg.1 NR_038855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21004138.hg.1 TC21000362.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100153157.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100153157.hg.1 NR_038855.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100153157.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100153157.hg.1 NR_038855.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100153157.hg.1 TC2100007880.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 ENST00000451410.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 NR_038855.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 NR_038855.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007880.hg.1 NR_038855.1 Refseq
          Human Genome U133 Plus 2.0 Array 1563385_at - -
          Human Genome U133 Plus 2.0 Array 1563385_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563385_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018738 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018738 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 131079 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1858963 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_114601 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_114601 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1858963 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1858963 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1858963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline