Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00417248_m1
          See other HCG22 GE Assays ›
          Gene Symbol:
          HCG22
          Gene Name:
          HLA complex group 22
          Gene Aliases:
          PBMUCL2
          Chromosome Location:
          Chr.6: 2535826 - 2542227 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00417248_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HCG22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003948.3 3 - 4 1697 69
          NR_145427.2 3 - 4 1189 69
          GenBank mRNA AB560771.1 - 3 - 4 1184 69
          AB600644.1 - 3 - 4 1692 69
          AK094433.1 - 2 - 3 1508 69

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642264 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04642238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24833782 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 583283 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 433954 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3242453 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P859499 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 20633 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13247 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3242453 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3242453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833802 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833802 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04642229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24833798 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P859499 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P859499 - -
          Agilent Human 1B Microarray G4111A A_32_P874469 - -
          Agilent Human 1B Microarray G4111A A_32_P859499 - -
          Agilent Human 1B Microarray G4111A A_32_P301783 - -
          Agilent Human 1B Microarray G4111A A_32_P346593 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642286 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642269 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642295 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642218 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642291 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642297 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642284 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642310 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642231 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642265 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15799721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15799725 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642321 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04642293 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17496598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560767_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560767_at - -
          Human Genome U133 Plus 2.0 Array 1560767_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560767_at2 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031267 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031267 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230765 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1667229 AK094433.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1667229 NR_003948.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1667229 AB560771.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667229 AK094433.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667229 NR_003948.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667229 AB560771.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1667229 AK094433.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1667229 NR_003948.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1667229 AB560771.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1667229 AK094433.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1667229 NR_003948.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1667229 AB560771.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1667229 AK094433.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1667229 NR_003948.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1667229 AB560771.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline