Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00418633_m1
          See other SF3B3 GE Assays ›
          Gene Symbol:
          SF3B3
          Gene Name:
          splicing factor 3b subunit 3
          Gene Aliases:
          RSE1, SAP130, SF3b130, STAF130
          Chromosome Location:
          Chr.16: 70523816 - 70577668 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00418633_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SF3B3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012426.5 NP_036558.3 18 - 19 2640 59
          GenBank mRNA AJ001443.1 - 18 - 19 2694 59
          AK001297.1 - 7 - 8 1047 59
          AK074933.1 - 12 - 13 1563 59
          AK291768.1 - 18 - 19 2666 59
          BC068974.1 - 18 - 19 2605 59
          D13642.1 - 1 - 2 139 59
          D87686.1 - 18 - 19 2613 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008275.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000579.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SF3B3

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          regulation of DNA repair
          mRNA processing
          RNA splicing
          negative regulation of protein catabolic process
          regulation of RNA splicing
          positive regulation of transcription, DNA-templated
          U2-type prespliceosome assembly
          nucleic acid binding
          protein binding
          U2 snRNA binding
          macromolecular complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000302516.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000562874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000563739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000564899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000565990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000566095.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000567654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000568539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000569687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000572365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 ENST00000577085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008275.hg.1 NM_012426.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline