Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00610256_g1
          See other DUSP1 GE Assays ›
          Gene Symbol:
          DUSP1
          Gene Name:
          dual specificity phosphatase 1
          Gene Aliases:
          CL100, HVH1, MKP-1, MKP1, PTPN10
          Chromosome Location:
          Chr.5: 172768096 - 172771195 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00610256_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DUSP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004417.4 NP_004408.1 2 - 3 758 63
          GenBank mRNA AK298047.1 - 3 - 4 697 63
          AK298658.1 - 3 - 4 563 63
          AK299391.1 - 3 - 4 767 63
          AK300487.1 - 3 - 4 677 63
          AK304475.1 - 3 - 4 555 63
          AK316321.1 - 3 - 4 506 63
          BC022463.1 - 2 - 3 758 63
          X68277.1 - 2 - 3 748 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500012842.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05002066.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Human MAP Kinase Pathways Human Antioxident Mechanisms Inventoried Human_Map_Kinase Human_Antioxident_Mechanisms Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DUSP1

          Gene Ontology Categories:

          Function(s) Process(es)

          endoderm formation
          negative regulation of cell adhesion
          signal transduction
          negative regulation of cell proliferation
          response to light stimulus
          response to estradiol
          response to retinoic acid
          cellular response to hormone stimulus
          response to testosterone
          peptidyl-tyrosine dephosphorylation
          intracellular signal transduction
          peptidyl-threonine dephosphorylation
          response to hydrogen peroxide
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          response to glucocorticoid
          negative regulation of meiotic cell cycle
          response to cAMP
          response to calcium ion
          peptidyl-serine dephosphorylation
          ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of monocyte chemotaxis
          regulation of mitotic cell cycle spindle assembly checkpoint
          negative regulation of p38MAPK cascade
          cellular response to chemokine
          negative regulation of DNA biosynthetic process
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity
          hydrolase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          growth factor binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05028434.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05028434.hg.1 ENST00000434080.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05028434.hg.1 ENST00000457103.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05028434.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05028434.hg.1 TC05002066.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay TC0500012842.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500189332.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500189332.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500189332.hg.1 ENST00000239223.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500189332.hg.1 NM_004417.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500189332.hg.1 TC0500012842.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline