Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00744410_s1
          See other LOC541472 GE Assays ›
          Gene Symbol:
          LOC541472
          Gene Name:
          uncharacterized LOC541472
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 22725395 - 22727620 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00744410_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC541472
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_131935.1 2 1214 99
          GenBank mRNA BC027978.1 - 2 1220 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010449.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P305818 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P67060 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4539 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17291 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27358 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3249674 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P305818 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P67060 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P67060 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P305818 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 9129 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 8452 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 10181 - -
          Agilent Human 1B Microarray G4111A A_32_P67060 - -
          Agilent Human 1B Microarray G4111A A_32_P189757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8138544 - -
          Affymetrix Human Gene 1.0 ST Array 8138543 - -
          Affymetrix HT HG-U133+ PM Array Plate 243977_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0700010449.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010449.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010449.hg.1 NR_131935.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010449.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010449.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700010449.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700010449.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010449.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174330.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174330.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174330.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174330.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174330.hg.1 TC0700010449.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700174329.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174329.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174329.hg.1 ENST00000325042.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174329.hg.1 NR_131935.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174329.hg.1 TC0700010449.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 243977_PM_at - -
          Human Genome U133 Plus 2.0 Array 243977_at - -
          Human Genome U95 Set 59860_PM_at - -
          Human Genome U95 Set 59860_at - -
          Human Genome U133 Set 243977_at - -
          Human Genome U133 Set 243977_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 223324 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3288752 BC027978.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline