Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00851863_s1
          See other H3C7 GE Assays ›
          Gene Symbol:
          H3C7
          Gene Name:
          H3 clustered histone 7
          Gene Aliases:
          H3/i, H3C1, H3C10, H3C11, H3C12, H3C2, H3C3, H3C4, H3C6, H3C8, H3FI, HIST1H3F
          Chromosome Location:
          Chr.6: 26250142 - 26250635 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00851863_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H3C7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021018.3 NP_066298.1 1 458 65
          GenBank mRNA BC067492.1 - 1 428 65
          BC069818.1 - 1 430 65
          BC096131.2 - 1 428 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011140.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001353.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H3C7

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          nucleosome assembly
          gene expression
          telomere organization
          regulation of gene expression, epigenetic
          DNA binding
          protein binding
          structural constituent of chromatin
          cadherin binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24813755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24813754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04625494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04625495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24813753 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P30799 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P30799 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 23398 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 23605 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 29433 - -
          Agilent Human Genome CGH Microarray 44B A_14_P138808 - -
          Agilent Human Genome CGH Microarray 44A A_14_P138808 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52579 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P30799 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P30799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24813754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24813754 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P30799 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P138808 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625495 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625490 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625491 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625493 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625500 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625497 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625501 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625491 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625501 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625490 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625489 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P138808 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P97914 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P8004 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P30799 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 144 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 3128 - -
          Agilent Human 1B Microarray G4111A A_32_P206109 - -
          Agilent Human 1B Microarray G4111A A_32_P301330 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625513 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625508 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625469 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625507 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625510 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625442 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625505 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625480 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625471 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625462 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625456 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625451 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625449 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625493 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625464 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625475 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625491 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_021018_at - -
          Affymetrix Human Gene 1.0 ST Array 8124438 - -
          Affymetrix Human Gene 1.0 ST Array 8124439 - -
          Affymetrix Human Gene 1.1 ST Array 8124437 - -
          Affymetrix HT HG-U133+ PM Array Plate 208506_PM_at - -
          Affymetrix Human Genome U219 Array 11734574_x_at - -
          Affymetrix Human Genome U219 Array 11734573_at - -
          Affymetrix Human Genome U219 Array 11734574_PM_x_at - -
          Affymetrix Human Genome U219 Array 11734573_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016173.hg.1 ENST00000446824.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016173.hg.1 NM_021018.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016173.hg.1 TC06001353.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600175358.hg.1 NM_021018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175358.hg.1 NM_021018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175358.hg.1 TC0600011140.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 208575_PM_at - -
          Human Genome U133 Plus 2.0 Array 208496_x_at - -
          Human Genome U133 Plus 2.0 Array 214472_at - -
          Human Genome U133 Plus 2.0 Array 208506_at - -
          Human Genome U133 Plus 2.0 Array 208577_PM_at - -
          Human Genome U133 Plus 2.0 Array 214616_PM_at - -
          Human Genome U133 Plus 2.0 Array 214472_PM_at - -
          Human Genome U133 Plus 2.0 Array 208577_at - -
          Human Genome U133 Plus 2.0 Array 208496_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 214616_at - -
          Human Genome U133 Plus 2.0 Array 208506_PM_at - -
          Human Genome U133 Plus 2.0 Array 208575_at - -
          Human Genome U133 Plus 2.0 Array 208576_s_at - -
          Human Genome U133 Plus 2.0 Array 208576_PM_s_at - -
          Human Genome U133A 2.0 Array 208577_at - -
          Human Genome U133A 2.0 Array 208496_PM_x_at - -
          Human Genome U133A 2.0 Array 208577_PM_at - -
          Human Genome U133A 2.0 Array 214472_PM_at - -
          Human Genome U133A 2.0 Array 208575_PM_at - -
          Human Genome U133A 2.0 Array 208575_at - -
          Human Genome U133A 2.0 Array 214616_PM_at - -
          Human Genome U133A 2.0 Array 214472_at - -
          Human Genome U133A 2.0 Array 208506_at - -
          Human Genome U133A 2.0 Array 208576_PM_s_at - -
          Human Genome U133A 2.0 Array 208576_s_at - -
          Human Genome U133A 2.0 Array 208506_PM_at - -
          Human Genome U133A 2.0 Array 214616_at - -
          Human Genome U133A 2.0 Array 208496_x_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_021018_at - -
          AffymetrixPathwork Diagnostics Pathchip pw534358 - -
          Human Genome U95 Set 35598_at - -
          Human Genome U95 Set 35598_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208506_at2 - -
          Human Genome U133 Set 214616_PM_at - -
          Human Genome U133 Set 208575_PM_at - -
          Human Genome U133 Set 208506_PM_at - -
          Human Genome U133 Set 208577_at - -
          Human Genome U133 Set 208577_PM_at - -
          Human Genome U133 Set 214616_at - -
          Human Genome U133 Set 208576_PM_s_at - -
          Human Genome U133 Set 208496_x_at - -
          Human Genome U133 Set 208575_at - -
          Human Genome U133 Set 208506_at - -
          Human Genome U133 Set 214472_PM_at - -
          Human Genome U133 Set 208576_s_at - -
          Human Genome U133 Set 214472_at - -
          Human Genome U133 Set 208496_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029538 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029538 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1788489 BC067492.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1788489 BC069818.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1788489 BC096131.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1788489 CR541983.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396497-S BC067492.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396497-S BC069818.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396497-S BC096131.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396497-S NM_021018.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_8533-S - -
          Illumina human-6 v1.0 expression beadchip 0005910408 - -
          Illumina humanRef-8 v1.0 expression beadchip 0005910408 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2030402 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_8533 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1788489 CR542011.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1788489 BC067492.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1788489 BC069818.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1788489 BC096131.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1788489 CR541983.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1788489 CR542011.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0