Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00863947_g1
          See other LINC00547 GE Assays ›
          Gene Symbol:
          LINC00547
          Gene Name:
          long intergenic non-protein coding RNA 547
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 37534940 - 37551536 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00863947_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00547
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040244.1 4 - 5 271 105
          GenBank mRNA AK092066.1 - 4 - 5 271 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300006901.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000129.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786321 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786265 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16627225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16627222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786322 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16627227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08786257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29233094 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111345 AF089841.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111345 AJ012737.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 795312 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 301308 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 927288 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 713164 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 653597 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 566454 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 20034 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 AF039568.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 AK074576.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 AK292856.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 BC006178.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 BC018437.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR595143.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR597095.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR600018.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR600923.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR606001.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR615911.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR621586.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 CR749670.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 NM_004206.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 203562 NM_032970.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 332717 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 317804 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 222064 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 86387 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 242827 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 64679 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 253816 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 90162 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3652730 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P39918007 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P39918007 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3652730 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3652730 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P39918007 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P39918007 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19779833 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 172286 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 17151 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39918007 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19779833 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P19617462 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P19617790 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39917979 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39918007 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19779833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13001487.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001487.hg.1 TC13000129.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146914.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146914.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146914.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146914.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146914.hg.1 TC1300006901.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011346.hg.1 ENST00000354527.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146913.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146913.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146913.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146913.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146913.hg.1 TC1300006901.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300146915.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146915.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146915.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300146915.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300146915.hg.1 TC1300006901.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 ENST00000612438.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006901.hg.1 NR_040244.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050070 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050070 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 680477 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline