| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000281701.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000340871.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000361463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000391875.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000469075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
ENST00000482491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
NM_001243146.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
NM_001243147.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
NM_002533.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
NM_206840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059096.hg.1 |
TC01003881.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000281701.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000340871.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000391875.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000469075.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000469968.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000482491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_001243146.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_001243147.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_002533.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_206840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000281701.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000340871.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000391875.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000469075.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000469968.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
ENST00000482491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_001243146.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_001243147.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_002533.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
NM_206840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232132.hg.1 |
TC0100017467.hg.1 |
Clariom D TC |