Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00968671_m1
          See other HTR2C GE Assays ›
          Gene Symbol:
          HTR2C
          Gene Name:
          5-hydroxytryptamine receptor 2C
          Gene Aliases:
          5-HT1C, 5-HT2C, 5-HTR2C, 5HTR2C, HTR1C
          Chromosome Location:
          Chr.X: 114584078 - 114910061 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00968671_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HTR2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000868.3 NP_000859.1 4 - 5 1067 118
          NM_001256760.2 NP_001243689.1 5 - 6 1158 118
          NM_001256761.2 NP_001243690.1 4 - 5 1067 118
          GenBank mRNA AF208053.1 - 2 - 3 339 118
          AF498983.1 - 2 - 3 339 118
          AK295753.1 - 4 - 5 1059 118
          AK308766.1 - 4 - 5 646 118
          BC028688.1 - 5 - 6 1020 118
          BC095543.1 - 2 - 3 340 6519218 118
          FJ200493.1 - 2 - 3 339 118
          M81778.1 - 5 - 6 1126 118
          U49516.1 - 4 - 5 1067 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008178.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000555.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HTR2C

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          cGMP biosynthetic process
          cellular calcium ion homeostasis
          phospholipase C-activating G-protein coupled receptor signaling pathway
          phospholipase C-activating serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          chemical synaptic transmission
          locomotory behavior
          feeding behavior
          positive regulation of phosphatidylinositol biosynthetic process
          regulation of neurological system process
          regulation of appetite
          response to drug
          regulation of corticotropin-releasing hormone secretion
          positive regulation of fat cell differentiation
          release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          Gq/11-coupled serotonin receptor activity
          G-protein coupled serotonin receptor activity
          drug binding
          serotonin binding
          1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X007422.hg.1 TC0X000555.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X007422.hg.1 ENST00000276198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007422.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007422.hg.1 ENST00000371951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007422.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00008178.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000276198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000371950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 ENST00000371951.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_000868.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_001256760.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 NM_001256761.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155825.hg.1 TC0X00008178.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline