Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01007306_m1
          See other PRAMENP GE Assays ›
          Gene Symbol:
          PRAMENP
          Gene Name:
          PRAME N-terminal like, pseudogene
          Gene Aliases:
          PRAMEF24P, PRAMEL
          Chromosome Location:
          Chr.22: 21991099 - 22043934 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01007306_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRAMENP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_135291.1 6 - 7 2381 82
          GenBank mRNA AK131325.1 - 6 - 7 2381 82

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11438023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32113654 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P273180 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5524 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 33263 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P273180 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P273180 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03603698 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21286946 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03603672 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03603698 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21286946 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03603726 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21286992 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03603672 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03603687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8074841 - -
          Affymetrix Human Gene 1.0 ST Array 8074840 - -
          Affymetrix Human Gene 1.0 ST Array 8074839 - -
          Affymetrix Human Gene 1.0 ST Array 8074837 - -
          Affymetrix Human Gene 1.0 ST Array 8074838 - -
          Affymetrix Human Gene 1.1 ST Array 8074836 - -
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000337471.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000419303.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008113.hg.1 ENST00000548391.1 Ensembl
          Human Genome U95 Set 57448_at - -
          Human Genome U95 Set 57448_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739609 AK131325.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1787834 AK131325.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1739609 AK131325.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1739609 AK131325.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1787834 AK131325.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1739609 AK131325.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline