Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01071998_m1
          See other KHK GE Assays ›
          Gene Symbol:
          KHK
          Gene Name:
          ketohexokinase
          Gene Aliases:
          FRUCTU
          Chromosome Location:
          Chr.2: 27086772 - 27100762 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01071998_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KHK
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000221.3 NP_000212.1 4 - 5 904 72
          NM_006488.3 NP_006479.1 4 - 5 904 72
          XM_005264294.5 XP_005264351.1 5 - 6 1042 72
          XM_005264296.5 XP_005264353.1 4 - 5 907 72
          XM_005264298.5 XP_005264355.1 2 - 3 652 72
          XM_006712008.5 XP_006712071.1 5 - 6 1042 72
          XM_006712009.5 XP_006712072.1 5 - 6 1039 72
          XM_006712010.5 XP_006712073.1 4 - 5 907 72
          XM_006712011.5 XP_006712074.1 4 - 5 904 72
          XM_006712012.5 XP_006712075.1 4 - 5 904 72
          XM_006712013.5 XP_006712076.1 3 - 4 769 72
          XM_006712014.5 XP_006712077.1 2 - 3 652 72
          XM_017004060.3 XP_016859549.1 5 - 6 1039 72
          XM_017004061.3 XP_016859550.1 3 - 4 769 72
          GenBank mRNA AK130033.1 - 4 - 5 450 72
          AK223075.1 - 4 - 5 450 72
          BC006233.2 - 4 - 5 871 72
          BX648873.1 - 2 - 3 200 72
          CR456801.1 - 4 - 5 416 72
          HM005415.1 - 4 - 5 933 72
          X78677.1 - 2 - 3 212 72
          X78678.1 - 4 - 5 424 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200007053.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000164.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KHK

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose metabolic process
          response to sucrose
          response to glucose
          response to fructose
          response to zinc ion
          response to insulin
          carbohydrate phosphorylation
          regulation of glycogen metabolic process
          nucleotide binding
          ketohexokinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          identical protein binding
          protein homodimerization activity
          fructose binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260598.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000260599.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000429697.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000464371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000469936.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000469936.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000469936.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000469936.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 ENST00000490823.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_000221.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007053.hg.1 NM_006488.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline