Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01106214_m1
          See other PTPN13 GE Assays ›
          Gene Symbol:
          PTPN13
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 13
          Gene Aliases:
          FAP-1, PNP1, PTP-BAS, PTP-BL, PTP1E, PTPL1, PTPLE, hPTP1E
          Chromosome Location:
          Chr.4: 86594307 - 86815179 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01106214_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTPN13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006264.2 NP_006255.1 29 - 30 5221 65
          NM_080683.2 NP_542414.1 30 - 31 5278 65
          NM_080684.2 NP_542415.1 27 - 28 4705 65
          NM_080685.2 NP_542416.1 30 - 31 5293 65
          XM_011532165.1 XP_011530467.1 30 - 31 5301 65
          XM_017008511.1 XP_016864000.1 30 - 31 5286 65
          XM_017008512.1 XP_016864001.1 29 - 30 5244 65
          XM_017008513.1 XP_016864002.1 29 - 30 5229 65
          XM_017008514.1 XP_016864003.1 29 - 30 5244 65
          XM_017008515.1 XP_016864004.1 29 - 30 4740 65
          XM_017008516.1 XP_016864005.1 27 - 28 4728 65
          GenBank mRNA AB208904.1 - 28 - 29 4645 65
          AF233323.1 - 7 - 8 832 65
          BC039610.1 - 7 - 8 832 65
          BC140777.1 - 28 - 29 4760 9021645 65
          D21209.1 - 30 - 31 4861 65
          D21210.1 - 29 - 30 4804 65
          D21211.1 - 27 - 28 4288 65
          L34583.1 - 7 - 8 979 65
          U12128.1 - 29 - 30 5030 65
          X79676.1 - 7 - 8 1151 65
          X80289.1 - 29 - 30 4818 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400008041.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000469.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTPN13

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine dephosphorylation
          protein tyrosine phosphatase activity
          protein binding
          phosphatidylinositol 3-kinase regulatory subunit binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 TC04000469.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000316707.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000357349.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000411767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000427191.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000436978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 ENST00000511467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 NM_006264.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 NM_080683.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 NM_080684.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04006753.hg.1 NM_080685.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000316707.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000411767.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000427191.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000436978.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000511467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_006264.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080683.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080684.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080685.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000316707.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000411767.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000427191.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000436978.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 ENST00000511467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_006264.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080683.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080684.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 NM_080685.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400155353.hg.1 TC0400008041.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 188871 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline