Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01115614_m1
          See other ABCD2 GE Assays ›
          Gene Symbol:
          ABCD2
          Gene Name:
          ATP binding cassette subfamily D member 2
          Gene Aliases:
          ABC39, ALDL1, ALDR, ALDRP, hALDR
          Chromosome Location:
          Chr.12: 39530966 - 39651333 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01115614_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ABCD2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005164.3 NP_005155.1 1 - 2 1364 114
          XM_011538027.2 XP_011536329.1 2 - 3 1304 114
          XM_017018992.1 XP_016874481.1 1 - 2 1364 114
          XM_017018993.1 XP_016874482.1 1 - 2 1364 114
          XM_017018994.1 XP_016874483.1 1 - 2 1364 114
          GenBank mRNA AJ000327.1 - 1 - 2 1073 114
          AK314254.1 - 1 - 2 1364 114
          BC104901.1 - 1 - 2 970 8143904 114
          BC104903.1 - 1 - 2 964 8143906 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010413.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001392.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ABCD2

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          fatty acid beta-oxidation
          long-chain fatty acid transport
          positive regulation of fatty acid beta-oxidation
          very long-chain fatty acid catabolic process
          transmembrane transport
          long-chain fatty acid transporter activity
          protein binding
          ATP binding
          ATPase activity, coupled to transmembrane movement of substances
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018075.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12018075.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018075.hg.1 TC12001392.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12018076.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12018076.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018076.hg.1 TC12001392.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12018078.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12018078.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018078.hg.1 TC12001392.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12018079.hg.1 NM_005164.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018079.hg.1 TC12001392.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200178569.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178569.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178569.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178569.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178569.hg.1 TC1200010413.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178570.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178570.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178570.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178570.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178570.hg.1 TC1200010413.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200178573.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178573.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178573.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178573.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178573.hg.1 TC1200010413.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200178571.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178571.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178571.hg.1 ENST00000308666.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178571.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178571.hg.1 TC1200010413.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200178574.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178574.hg.1 NM_005164.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178574.hg.1 TC1200010413.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline