| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028966.hg.1 |
ENST00000432045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028966.hg.1 |
NR_034120.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028966.hg.1 |
TC07001868.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
NR_110473.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
NR_110473.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190944.hg.1 |
TC0700012613.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000418546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000447307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000447307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_034120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_110472.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_110473.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000418546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000432045.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000447307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
ENST00000447307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_034120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_110472.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
NR_110473.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700190961.hg.1 |
TC0700012613.hg.1 |
Clariom D TC |