Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01555465_m1
          See other ATG5 GE Assays ›
          Gene Symbol:
          ATG5
          Gene Name:
          autophagy related 5
          Gene Aliases:
          APG5, APG5-LIKE, APG5L, ASP, hAPG5
          Chromosome Location:
          Chr.6: 106184476 - 106325820 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01555465_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATG5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286107.1 NP_001273036.1 1 - 2 296 95
          NM_001286108.1 NP_001273037.1 1 - 2 296 95
          NM_001286111.1 NP_001273040.1 1 - 2 296 95
          NM_004849.3 NP_004840.1 1 - 2 296 95
          GenBank mRNA AK001899.1 - 1 - 2 263 95
          AK299577.1 - 1 - 2 288 95
          AK315800.1 - 1 - 2 134 95
          BC093011.1 - 1 - 2 223 30345897 95
          BP297166.1 - 1 - 2 296 95
          BX537904.1 - 1 - 2 133 95
          CR749386.1 - 1 - 2 215 95
          Y11588.1 - 1 - 2 269 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012731.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001982.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK001899.1,AK299577.1,BC093011.1,BX537904.1,CR749386.1,NM_001286107.1,NM_001286108.1,NM_001286111.1,NM_004849.3,Y11588.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATG5

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          blood vessel remodeling
          regulation of cytokine secretion involved in immune response
          C-terminal protein lipidation
          autophagy
          apoptotic process
          cellular response to nitrogen starvation
          response to fungus
          macroautophagy
          cellular homeostasis
          antigen processing and presentation of endogenous antigen
          negative regulation of protein ubiquitination
          aggrephagy
          negative stranded viral RNA replication
          vasodilation
          response to drug
          negative regulation of apoptotic process
          post-translational protein modification
          nucleophagy
          negative thymic T cell selection
          innate immune response
          otolith development
          regulation of release of sequestered calcium ion into cytosol
          ventricular cardiac muscle cell development
          heart contraction
          negative regulation of cell death
          protein lipidation involved in autophagosome assembly
          positive regulation of mucus secretion
          regulation of cilium assembly
          positive regulation of mitophagy
          negative regulation of reactive oxygen species metabolic process
          negative regulation of histone H4-K16 acetylation
          protein binding
          Atg8 ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0