Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01573505_m1
          See other WNT4 GE Assays ›
          Gene Symbol:
          WNT4
          Gene Name:
          Wnt family member 4
          Gene Aliases:
          SERKAL, WNT-4
          Chromosome Location:
          Chr.1: 22117313 - 22143097 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01573505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WNT4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030761.5 NP_110388.2 2 - 3 485 66
          XM_011541597.3 XP_011539899.1 2 - 3 471 66
          GenBank mRNA AB061675.1 - 2 - 3 315 66
          AF316543.1 - 2 - 3 354 66
          AK296058.1 - 2 - 3 394 66
          AY358947.1 - 2 - 3 346 66
          BC057781.1 - 2 - 3 505 66
          BT020125.1 - 2 - 3 310 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013246.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002330.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WNT4

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          branching involved in ureteric bud morphogenesis
          kidney development
          mesonephros development
          epithelial to mesenchymal transition
          embryonic epithelial tube formation
          liver development
          gamete generation
          sex differentiation
          fibroblast growth factor receptor signaling pathway
          male gonad development
          female gonad development
          cellular response to starvation
          negative regulation of gene expression
          negative regulation of steroid biosynthetic process
          Wnt signaling pathway
          regulation of cell-cell adhesion
          cell differentiation
          neuron differentiation
          female sex determination
          adrenal gland development
          negative regulation of cell migration
          positive regulation of bone mineralization
          positive regulation of aldosterone biosynthetic process
          positive regulation of collagen biosynthetic process
          T cell differentiation in thymus
          immature T cell proliferation in thymus
          tube morphogenesis
          non-canonical Wnt signaling pathway
          negative regulation of fibroblast growth factor receptor signaling pathway
          hormone metabolic process
          positive regulation of MAPK cascade
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of Ras protein signal transduction
          oocyte development
          branching morphogenesis of an epithelial tube
          smooth muscle cell differentiation
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          Sertoli cell differentiation
          canonical Wnt signaling pathway
          somatotropin secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          mesenchymal to epithelial transition
          tertiary branching involved in mammary gland duct morphogenesis
          kidney morphogenesis
          negative regulation of wound healing
          mammary gland epithelium development
          positive regulation of dermatome development
          paramesonephric duct development
          negative regulation of testicular blood vessel morphogenesis
          cellular response to transforming growth factor beta stimulus
          nephron development
          renal vesicle formation
          renal vesicle induction
          metanephric mesenchymal cell differentiation
          mesonephric tubule development
          metanephric tubule formation
          metanephric nephron development
          metanephric nephron morphogenesis
          apoptotic signaling pathway
          meiotic nuclear division
          pericyte cell differentiation
          negative regulation of male gonad development
          positive regulation of cortisol biosynthetic process
          negative regulation of androgen biosynthetic process
          negative regulation of testosterone biosynthetic process
          negative regulation of apoptotic signaling pathway
          transcription corepressor activity
          receptor binding
          frizzled binding
          cytokine activity
          receptor agonist activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01035973.hg.1 ENST00000290167.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035973.hg.1 ENST00000542383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035973.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035973.hg.1 TC01002330.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000415567.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000415567.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000441048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 ENST00000441048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000415567.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000415567.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000441048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 ENST00000441048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013246.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198253.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198253.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198253.hg.1 ENST00000290167.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198253.hg.1 NM_030761.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198253.hg.1 TC0100013246.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline