Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01653052_mH
          See other FOXI2 GE Assays ›
          Gene Symbol:
          FOXI2
          Gene Name:
          forkhead box I2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 127737274 - 127741186 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01653052_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXI2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207426.2 NP_997309.2 1 - 2 504 169
          GenBank mRNA AK128865.1 - 1 - 2 90 169

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009287.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000921.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK128865.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FOXI2

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10011333.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10011333.hg.1 NM_207426.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10011333.hg.1 TC10000921.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10011334.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10011334.hg.1 NM_207426.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10011334.hg.1 TC10000921.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay TC1000009287.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163657.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163657.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163657.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163657.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163657.hg.1 TC1000009287.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163658.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163658.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163658.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163658.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163658.hg.1 TC1000009287.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000163659.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163659.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163659.hg.1 ENST00000388920.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000163659.hg.1 NM_207426.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000163659.hg.1 TC1000009287.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline