Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01881698_s1
          See other P2RY12 GE Assays ›
          Gene Symbol:
          P2RY12
          Gene Name:
          purinergic receptor P2Y12
          Gene Aliases:
          ADPG-R, BDPLT8, HORK3, P2T(AC), P2Y(12)R, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12, SP1999
          Chromosome Location:
          Chr.3: 151336843 - 151384812 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01881698_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2RY12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022788.4 NP_073625.1 3 1215 136
          NM_176876.2 NP_795345.1 2 1189 136
          XM_017007069.1 XP_016862558.1 3 1473 136
          GenBank mRNA AB052684.1 - 1 915 136
          AF313449.1 - 2 989 136
          AF321815.1 - 2 986 136
          AJ320495.1 - 1 932 136
          AK292096.1 - 3 1154 136
          AK303714.1 - 4 865 136
          AK316575.1 - 3 1157 136
          AY136754.1 - 1 915 136
          BC017898.1 - 3 1159 4251263 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300012816.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001907.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2RY12

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          adenosine receptor signaling pathway
          substrate-dependent cell migration, cell extension
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          hemostasis
          glial cell migration
          negative regulation of norepinephrine secretion
          G-protein coupled purinergic nucleotide receptor signaling pathway
          positive regulation of ion transport
          protein kinase B signaling
          positive regulation of GTPase activity
          negative regulation of cell differentiation
          regulation of calcium ion transport
          platelet aggregation
          calcium ion transmembrane transport
          cellular response to ATP
          potassium ion transmembrane transport
          regulation of microglial cell migration
          G-protein coupled adenosine receptor activity
          ADP receptor activity
          guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034363.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034363.hg.1 ENST00000455408.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034363.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034363.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034363.hg.1 TC03001907.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03034364.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034364.hg.1 ENST00000455408.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03034364.hg.1 NM_022788.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034364.hg.1 NM_176876.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03034364.hg.1 TC03001907.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 ENST00000468596.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay TC0300012816.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 ENST00000302632.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 NM_022788.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 NM_176876.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300195926.hg.1 TC0300012816.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline