Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01884672_g1
          See other LINC00314 GE Assays ›
          Gene Symbol:
          LINC00314
          Gene Name:
          long intergenic non-protein coding RNA 314
          Gene Aliases:
          C21orf94, NCRNA00314
          Chromosome Location:
          Chr.21: 28013363 - 28023209 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01884672_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00314
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027246.1 2 232 131
          GenBank mRNA AF427489.1 - 2 213 131
          BC126226.1 - 2 189 131
          BC126230.1 - 2 189 131
          CB049373.1 - 2 232 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006833.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11333819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11333813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31992893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17228437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11333817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31992948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11333818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11333820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31992947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31992972 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 97445 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 59841 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 355911 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 752911 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 654474 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 52228 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 36994 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40812 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 197137 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 194022 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P352990 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3368383 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5065 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5066 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 23441 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37656 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5826 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P121780 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P121780 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3368383 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P352990 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3368383 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P352990 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992947 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992944 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992866 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992948 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228437 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228422 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992874 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333817 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992977 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333813 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333818 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333816 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333810 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992911 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333820 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333819 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333821 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992973 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992972 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992976 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228340 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228450 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992969 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992893 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17228340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17228422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333818 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333813 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333817 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992944 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992976 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228422 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992911 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228340 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992866 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333810 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333820 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992973 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333819 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333821 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992969 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992947 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992893 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992874 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228450 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992854 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992948 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333816 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992977 - -
          Agilent Whole Chr array_18 19 21 A_17_P17228437 - -
          Agilent Whole Chr array_18 19 21 A_17_P11333818 - -
          Agilent Whole Chr array_18 19 21 A_17_P31992972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17228422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17228340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31992866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11333810 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P121780 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P352990 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 29607 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 71550 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13895511 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P121780 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11333815 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11333817 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11333820 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11333818 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P13895511 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P121780 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21225796 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8067837 - -
          Affymetrix Human Gene 1.0 ST Array 8068053 - -
          Affymetrix Human Gene 1.0 ST Array 8068055 - -
          Affymetrix Human Gene 1.0 ST Array 8022437 - -
          Affymetrix Human Gene 1.0 ST Array 8068056 - -
          Affymetrix Human Gene 1.0 ST Array 8067838 - -
          Affymetrix Human Gene 1.0 ST Array 8022438 - -
          Affymetrix Human Gene 1.0 ST Array 8068054 - -
          Affymetrix Human Gene 1.1 ST Array 8068052 - -
          Affymetrix Human Gene 1.1 ST Array 8067836 - -
          Affymetrix Human Gene 1.1 ST Array 8022436 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553519_PM_at - -
          Affymetrix Human Genome U219 Array 11746707_at - -
          Affymetrix Human Genome U219 Array 11746707_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006833.hg.1 NR_027246.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145992.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145992.hg.1 NR_027246.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145992.hg.1 TC2100006833.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145992.hg.1 ENST00000324988.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145992.hg.1 NR_027246.1 Refseq
          Human Genome U133 Plus 2.0 Array 1553519_at - -
          Human Genome U133 Plus 2.0 Array 1553519_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553519_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019122 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019122 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 222541 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1767601 AF427489.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1767601 BC126226.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1767601 BC126230.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1767601 CB049373.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1767601 NR_027246.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1767601 AF427489.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1767601 BC126226.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1767601 BC126230.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1767601 CB049373.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline