Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01943074_g1
          See other OR10G8 GE Assays ›
          Gene Symbol:
          OR10G8
          Gene Name:
          olfactory receptor family 10 subfamily G member 8
          Gene Aliases:
          OR11-274, OR11-282
          Chromosome Location:
          Chr.11: 124026798 - 124030634 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01943074_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR10G8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004464.2 NP_001004464.1 2 934 159
          GenBank mRNA BC136917.1 - 1 870 159
          BC136920.1 - 1 870 159

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009370.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR10G8

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16512087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16512085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16512080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089538 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P125204 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P125204 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22910 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 23826 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33071652 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P125204 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P125204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089539 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089541 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089541 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16512081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089539 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089543 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P125204 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P125204 - -
          Agilent Human 1B Microarray G4111A A_32_P383225 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089553 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089519 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089499 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089575 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089566 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089525 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16512105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16512101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089511 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089578 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089500 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089533 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16512107 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089516 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089523 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089562 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089527 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089548 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089528 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089570 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16512077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089507 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089558 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089505 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089542 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089534 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089556 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004464_at - -
          Affymetrix Human Gene 1.0 ST Array 7944800 - -
          Affymetrix Human Gene 1.0 ST Array 7944802 - -
          Affymetrix Human Gene 1.1 ST Array 7944799 - -
          Affymetrix Human Gene 1.1 ST Array 7944801 - -
          Affymetrix Human Genome U219 Array 11759279_PM_x_at - -
          Affymetrix Human Genome U219 Array 11759279_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009370.hg.1 NM_001004464.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009370.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009370.hg.1 NM_001004464.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100170231.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170231.hg.1 ENST00000431524.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170231.hg.1 TC1100009370.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047072 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047072 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1731717 BC136917.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1731717 BC136920.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_8319-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4290139 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_8319 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1731717 NM_001004464.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1731717 BC136917.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1731717 BC136920.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1731717 NM_001004464.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline