Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02379130_s1
          See other C3ORF56 GE Assays ›
          Gene Symbol:
          C3orf56
          Gene Name:
          chromosome 3 open reading frame 56
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 127193131 - 127198185 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02379130_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C3orf56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007534.2 NP_001007535.1 2 1624 161
          GenBank mRNA AK097460.1 - 2 1627 161

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300008695.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C3orf56

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30669 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3210009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8082349 - -
          Affymetrix Human Gene 1.0 ST Array 8082348 - -
          Affymetrix Human Gene 1.1 ST Array 8082347 - -
          Affymetrix HT HG-U133+ PM Array Plate 1562762_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161735.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161735.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161735.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161735.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161735.hg.1 TC0300008695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay TC0300008695.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161732.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161732.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161732.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161732.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161732.hg.1 TC0300008695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300161733.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161733.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161733.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161733.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161733.hg.1 TC0300008695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300161734.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161734.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161734.hg.1 ENST00000624688.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300161734.hg.1 NM_001007534.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300161734.hg.1 TC0300008695.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1562762_at - -
          Human Genome U133 Plus 2.0 Array 1562762_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562762_at11 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1794112 BC137155.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1794112 AK097460.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1794112 BC137154.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1794112 BC137155.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1794112 AK097460.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1794112 BC137154.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline