Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02519820_s1
          See other LOC149950 GE Assays ›
          Gene Symbol:
          LOC149950
          Gene Name:
          uncharacterized LOC149950
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 32587479 - 32608893 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02519820_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC149950
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034152.1 3 460 81
          GenBank mRNA BC111383.1 - 3 460 5169644 81
          BC130462.1 - 2 258 40146493 81
          BC130464.1 - 2 258 40146495 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007125.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000214.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17187215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17187220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17187205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31781812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11134612 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43339 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3290235 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 36375 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506823 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506840 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506825 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34685783 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506826 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506837 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506832 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506843 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03506831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8061742 - -
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002646.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002646.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002646.hg.1 TC20000214.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002648.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002648.hg.1 ENST00000375671.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002648.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002648.hg.1 TC20000214.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002650.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002650.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002650.hg.1 TC20000214.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002647.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002647.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002647.hg.1 TC20000214.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002649.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002649.hg.1 ENST00000375671.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002649.hg.1 NR_034152.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002649.hg.1 TC20000214.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 TC2000007125.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148722.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 TC2000007125.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148724.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 TC2000007125.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148721.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148720.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148720.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148720.hg.1 TC2000007125.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148720.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148720.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 TC2000007125.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148723.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375670.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 ENST00000375671.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 NR_034152.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007125.hg.1 NR_034152.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1665740 BC130464.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1665740 BC130462.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1665740 NR_034152.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665740 BC111383.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665740 BC130464.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665740 BC130462.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665740 NR_034152.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665740 BC111383.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline