Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02576433_g1
          See other DEFB136 GE Assays ›
          Gene Symbol:
          DEFB136
          Gene Name:
          defensin beta 136
          Gene Aliases:
          DEFB137
          Chromosome Location:
          Chr.8: 1373422 - 1374084 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02576433_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFB136
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033018.2 NP_001028190.2 1 - 2 53 91
          GenBank mRNA AY621333.1 - 1 - 2 53 91
          DQ012026.1 - 1 - 2 53 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009625.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000981.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DEFB136

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          killing of cells of other organism
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          defense response to fungus
          antifungal innate immune response
          antibacterial innate immune response
          lipopolysaccharide binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16069037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16069035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959679 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16069039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16069036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16069029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959623 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3252598 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1192 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1191 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20854 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32207210 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P16383644 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3252598 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3252598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959685 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959685 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959667 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959636 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959681 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959672 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959644 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959685 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16069035 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959653 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959655 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959622 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959621 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959667 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959657 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959688 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959707 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959651 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959702 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959623 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959628 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959660 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959639 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959665 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959713 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959659 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959630 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959648 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959709 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959705 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959625 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959697 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959717 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959663 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959719 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16069039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959646 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959642 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001033018_at - -
          Affymetrix Human Gene 1.0 ST Array 8149348 - -
          Affymetrix Human Gene 1.0 ST Array 8149349 - -
          Affymetrix Human Gene 1.1 ST Array 8149347 - -
          Affymetrix Human Genome U219 Array 11735988_at - -
          Affymetrix Human Genome U219 Array 11735988_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0800009625.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009625.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009625.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009625.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012361.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012361.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012361.hg.1 TC08000981.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08012360.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08012360.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012360.hg.1 TC08000981.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800165486.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165486.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165486.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165486.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165486.hg.1 TC0800009625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800009625.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009625.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009625.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay TC0800009625.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165487.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165487.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165487.hg.1 ENST00000382209.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165487.hg.1 NM_001033018.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165487.hg.1 TC0800009625.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035528 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035528 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655553 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_22682-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1655553 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655553 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 870537 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_22682 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1778631 AY621333.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1655553 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1778631 AY621333.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline