Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03806006_s1
          See other RBMXL3 GE Assays ›
          Gene Symbol:
          RBMXL3
          Gene Name:
          RNA binding motif protein, X-linked like 3
          Gene Aliases:
          CXorf55
          Chromosome Location:
          Chr.X: 115189400 - 115192868 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03806006_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RBMXL3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145346.1 NP_001138818.1 1 2648 105
          GenBank mRNA AK097568.1 - 1 1671 105
          AK301904.1 - 1 2648 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008192.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000565.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P399864 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2031 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2032 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43419 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12764 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P399864 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P399864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631137 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631137 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11901064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32631160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17389078 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17389080 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901100 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901084 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901127 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901122 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901061 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901098 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17389067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901058 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901103 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901080 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901078 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901114 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11901066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17389064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8169464 - -
          Affymetrix Human Gene 1.1 ST Array 8169463 - -
          Affymetrix Human Genome U219 Array 11734279_at - -
          Affymetrix Human Genome U219 Array 11734280_x_at - -
          Affymetrix Human Genome U219 Array 11734280_PM_x_at - -
          Affymetrix Human Genome U219 Array 11734279_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR0X007438.hg.1 TC0X000565.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X007438.hg.1 ENST00000424776.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007438.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X007437.hg.1 TC0X000565.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X007437.hg.1 ENST00000424776.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007437.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008192.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008192.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155852.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155852.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155852.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155852.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155852.hg.1 TC0X00008192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00155851.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155851.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155851.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155851.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155851.hg.1 TC0X00008192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00008192.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008192.hg.1 ENST00000424776.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008192.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155849.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155849.hg.1 NM_001145346.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155849.hg.1 TC0X00008192.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041427 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041427 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225462 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686637 AK097568.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686637 AK301904.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686637 NM_001145346.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1686637 AK097568.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1686637 AK301904.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1686637 NM_001145346.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline