Search
Search
| Gene Symbol: JUND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286968.1 | NP_001273897.1 | 1 | 587 | 80 | |
| NM_005354.5 | NP_005345.3 | 1 | 587 | 80 | ||
| GenBank mRNA | AK223630.1 | - | 1 | 553 | 80 | |
| X51346.1 | - | 1 | 285 | 80 | ||
| X56681.1 | - | 1 | 589 | 80 | ||
Set Membership: |
|