Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04334126_m1
          See other TFPI2 GE Assays ›
          Gene Symbol:
          TFPI2
          Gene Name:
          tissue factor pathway inhibitor 2
          Gene Aliases:
          PP5, REF1, TFPI-2
          Chromosome Location:
          Chr.7: 93885397 - 93890991 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04334126_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TFPI2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271003.1 NP_001257932.1 2 - 3 552 66
          NM_001271004.1 NP_001257933.1 2 - 3 585 66
          NM_006528.3 NP_006519.1 2 - 3 585 66
          GenBank mRNA AK024817.1 - 2 - 3 347 66
          AK075427.1 - 2 - 3 346 66
          AK092499.1 - 2 - 3 314 66
          AK129833.1 - 2 - 3 347 66
          AK313260.1 - 2 - 3 347 66
          AY691946.1 - 2 - 3 323 66
          BC005330.1 - 2 - 3 347 3929933 66
          BC034155.1 - 1 - 2 70 66
          D29992.1 - 2 - 3 328 66
          DA833200.1 - 2 - 3 346 66
          L27624.1 - 2 - 3 310 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700011809.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TFPI2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          extracellular matrix structural constituent

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000222543.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000451238.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000461482.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_001271003.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_001271004.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_006528.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000222543.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000451238.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 ENST00000461482.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_001271003.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_001271004.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 NM_006528.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700184385.hg.1 TC0700011809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 ENST00000222543.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 ENST00000451238.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_001271003.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_001271004.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_006528.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 ENST00000222543.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 ENST00000451238.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_001271003.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_001271004.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 NM_006528.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700184382.hg.1 TC0700011809.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 144600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline