| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023936.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023936.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023936.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023936.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023936.hg.1 |
TC04001729.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023937.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023937.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023937.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023937.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023937.hg.1 |
TC04001729.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023935.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023935.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023935.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023935.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023935.hg.1 |
TC04001729.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023938.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023938.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023938.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023938.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023938.hg.1 |
TC04001729.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191389.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000502832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000507601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000502832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000507601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191393.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191388.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000502832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000502832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191392.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191390.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000361618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000393704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
ENST00000512698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001009554.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001301647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_001301648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
NM_021647.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191391.hg.1 |
TC0400012992.hg.1 |
Clariom D TC |