Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04935271_m1
          See other AIP GE Assays ›
          Gene Symbol:
          AIP
          Gene Name:
          aryl hydrocarbon receptor interacting protein
          Gene Aliases:
          ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2
          Chromosome Location:
          Chr.11: 67483029 - 67491108 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04935271_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001302959.1 NP_001289888.1 2 - 3 371 117
          NM_001302960.1 NP_001289889.1 2 - 3 405 117
          NM_003977.3 NP_003968.3 2 - 3 405 117
          GenBank mRNA BC104797.1 - 2 - 3 307 8143800 117
          BC104827.1 - 2 - 3 307 8143830 117
          BG821308.1 - 2 - 3 400 117
          BQ054206.1 - 2 - 3 371 117
          BQ420398.1 - 1 - 2 131 117
          U31913.1 - 2 - 3 284 117
          U78521.1 - 2 - 3 392 117

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AIP

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to mitochondrion
          xenobiotic metabolic process
          signal transduction
          regulation of protein kinase A signaling
          protein maturation by protein folding
          negative regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of nucleic acid-templated transcription
          transcription coactivator activity
          signal transducer activity
          protein binding
          transcription factor binding
          aryl hydrocarbon receptor binding
          GAF domain binding
          unfolded protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000279146.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000525341.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000528641.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 ENST00000529797.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302959.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_001302960.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008139.hg.1 NM_003977.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline