Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04997839_g1
          See other ANAPC2 GE Assays ›
          Gene Symbol:
          ANAPC2
          Gene Name:
          anaphase promoting complex subunit 2
          Gene Aliases:
          APC2
          Chromosome Location:
          Chr.9: 137174784 - 137188560 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04997839_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANAPC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013366.4 NP_037498.1 5 - 6 1199 100
          XM_047423274.1 XP_047279230.1 5 - 6 1199 100
          XM_047423275.1 XP_047279231.1 5 - 6 1199 100
          XM_047423276.1 XP_047279232.1 5 - 6 1199 100
          GenBank mRNA AB037827.1 - 3 - 4 420 100
          AF191337.1 - 5 - 6 1171 100
          AK294601.1 - 5 - 6 1199 100
          BC001579.1 - 3 - 4 436 100
          BC032503.1 - 5 - 6 1176 100

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANAPC2

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          metaphase/anaphase transition of mitotic cell cycle
          regulation of mitotic cell cycle
          nervous system development
          proteasomal protein catabolic process
          negative regulation of gene expression
          protein ubiquitination
          cell differentiation
          anaphase-promoting complex-dependent catabolic process
          positive regulation of synaptic plasticity
          positive regulation of axon extension
          positive regulation of dendrite morphogenesis
          cell division
          regulation of meiotic cell cycle
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          positive regulation of synapse maturation
          protein branched polyubiquitination
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000323927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000471131.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000483432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000485970.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000487917.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000493730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000495611.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 ENST00000618649.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq
          Affymetrix Human Clariom D Assay TC0900012039.hg.1 NM_013366.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-djsrz:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline