Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05021992_s1
          See other S1PR1 GE Assays ›
          Gene Symbol:
          S1PR1
          Gene Name:
          sphingosine-1-phosphate receptor 1
          Gene Aliases:
          CD363, CHEDG1, D1S3362, ECGF1, EDG-1, EDG1, S1P1
          Chromosome Location:
          Chr.1: 101236749 - 101241520 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05021992_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S1PR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320730.1 NP_001307659.1 2 1026 103
          NM_001400.4 NP_001391.2 2 1134 103
          GenBank mRNA AF022137.1 - 1 620 103
          AF233365.1 - 1 620 103
          AK297005.1 - 2 872 103
          AK312493.1 - 2 864 103
          BC018650.1 - 2 868 4071217 103
          CR541786.1 - 1 620 103
          CR542269.1 - 1 620 103
          M31210.1 - 2 870 103

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S1PR1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel maturation
          cardiac muscle tissue growth involved in heart morphogenesis
          sphingosine-1-phosphate signaling pathway
          chemotaxis
          cell adhesion
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          brain development
          cell migration
          transmission of nerve impulse
          lamellipodium assembly
          regulation of cell adhesion
          neuron differentiation
          positive regulation of cell migration
          regulation of bone mineralization
          actin cytoskeleton reorganization
          positive regulation of GTPase activity
          regulation of bone resorption
          endothelial cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle cell proliferation
          positive regulation of positive chemotaxis
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          negative regulation of stress fiber assembly
          heart trabecula morphogenesis
          T cell migration
          G-protein coupled receptor binding
          G-protein coupled receptor activity
          sphingosine-1-phosphate receptor activity
          sphingolipid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000305352.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 ENST00000475821.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009241.hg.1 NM_001400.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline