Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07292706_g1
          See other HLA-B GE Assays ›
          Gene Symbol:
          HLA-B BBX BCKDHA CCT6A CELF1 CEP41 CPSF6 FOSB FUT8 GCOM1 IFI16 LTF NOTCH3 PLEK PTGIS SBDS TCF4 TOP3B TRIM22 TYK2
          Gene Name:
          major histocompatibility complex, class I, B
          BBX high mobility group box domain containing
          branched chain keto acid dehydrogenase E1 subunit alpha
          chaperonin containing TCP1 subunit 6A
          CUGBP Elav-like family member 1
          centrosomal protein 41
          cleavage and polyadenylation specific factor 6
          FosB proto-oncogene, AP-1 transcription factor subunit
          fucosyltransferase 8
          GCOM1, MYZAP-POLR2M combined locus
          interferon gamma inducible protein 16
          lactotransferrin
          notch receptor 3
          pleckstrin
          prostaglandin I2 synthase
          SBDS ribosome maturation factor
          transcription factor 4
          DNA topoisomerase III beta
          tripartite motif containing 22
          tyrosine kinase 2
          Gene Aliases:
          AS, B-4901, HLAB
          ARTC1, HBP2, HSPC339, MDS001
          BCKDE1A, MSU, MSUD1, MSUD1A, OVD1A
          CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
          BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
          JBTS15, TSGA14
          CFIM, CFIM68, CFIM72, HPBRII-4, HPBRII-7
          AP-1, G0S3, GOS3, GOSB
          CDGF, CDGF1
          GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
          IFNGIP1, PYHIN2
          GIG12, HEL110, HLF2, LF
          CADASIL, CADASIL1, CASIL, IMF2, LMNS
          P47, PLEK1
          CYP8, CYP8A1, PGIS, PTGI
          CGI-97, SDO1, SDS, SWDS
          CDG2T, E2-2, FCD2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
          TOP3B1
          GPSTAF50, RNF94, STAF50
          IMD35, JTK1
          Chromosome Location:
          Chr.6: 2834235 - 2837538 on Build GRCh38
          Chromosome Location:
          Chr.3: 107522962 - 107811339 on Build GRCh38
          Chromosome Location:
          Chr.19: 41397818 - 41425002 on Build GRCh38
          Chromosome Location:
          Chr.7: 56051765 - 56063989 on Build GRCh38
          Chromosome Location:
          Chr.11: 47465937 - 47565539 on Build GRCh38
          Chromosome Location:
          Chr.7: 130393771 - 130441741 on Build GRCh38
          Chromosome Location:
          Chr.12: 69239569 - 69274358 on Build GRCh38
          Chromosome Location:
          Chr.19: 45467996 - 45475179 on Build GRCh38
          Chromosome Location:
          Chr.14: 65356842 - 65744121 on Build GRCh38
          Chromosome Location:
          Chr.15: 57591904 - 57717557 on Build GRCh38
          Chromosome Location:
          Chr.1: 158999976 - 159055151 on Build GRCh38
          Chromosome Location:
          Chr.3: 46435645 - 46485234 on Build GRCh38
          Chromosome Location:
          Chr.19: 15159038 - 15200995 on Build GRCh38
          Chromosome Location:
          Chr.2: 68365282 - 68397453 on Build GRCh38
          Chromosome Location:
          Chr.20: 49503874 - 49568137 on Build GRCh38
          Chromosome Location:
          Chr.7: 66987680 - 66995586 on Build GRCh38
          Chromosome Location:
          Chr.18: 55222185 - 55635957 on Build GRCh38
          Chromosome Location:
          Chr.22: 21957025 - 21982787 on Build GRCh38
          Chromosome Location:
          Chr.11: 5689790 - 5710863 on Build GRCh38
          Chromosome Location:
          Chr.19: 10350533 - 10380572 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Assay ID Hs07292706_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005514.8 NP_005505.2 4 850 66
          GenBank mRNA AB008102.1 - 4 829 66
          AF006634.1 - 4 829 66
          AF016641.1 - 4 829 66
          AF022783.1 - 4 829 66
          AF026218.1 - 4 829 66
          AF130734.1 - 5 829 66
          AF170577.1 - 4 829 66
          AF189017.1 - 4 829 66
          AF389378.1 - 4 833 66
          AF428252.1 - 4 833 66
          AJ131118.1 - 4 829 66
          AJ251593.1 - 4 828 66
          AJ535113.1 - 4 833 66
          AJ634528.1 - 4 829 66
          AK026595.1 - 5 852 66
          AK124160.1 - 4 1099 66
          AK129796.1 - 4 850 66
          AK130137.1 - 4 850 66
          AK130155.1 - 4 852 66
          AK130166.1 - 4 850 66
          AK130169.1 - 4 856 66
          AK130654.1 - 5 789 66
          AK290570.1 - 4 850 66
          AK292226.1 - 4 850 66
          AK300685.1 - 5 759 66
          AK313911.1 - 4 850 66
          AL576516.3 - 5 663 66
          AM711637.1 - 4 829 66
          AY007140.1 - 4 849 66
          AY102648.1 - 3 754 66
          AY569160.1 - 3 756 66
          BC007243.1 - 5 823 66
          BC013187.1 - 4 838 66
          BC091497.1 - 4 833 66
          BX647315.1 - 5 851 66
          D14343.1 - 4 793 66
          D25275.1 - 4 829 66
          D31816.1 - 4 829 66
          D49824.1 - 4 829 66
          D50290.1 - 4 829 66
          D50291.1 - 4 829 66
          D50292.1 - 4 829 66
          D50293.1 - 4 829 66
          D50294.1 - 4 829 66
          D50295.1 - 4 829 66
          D50296.1 - 4 829 66
          D50299.1 - 4 829 66
          D50300.1 - 4 829 66
          D83043.1 - 4 829 66
          D85761.1 - 5 829 66
          D85762.1 - 4 829 66
          D87665.1 - 5 820 66
          DQ336696.1 - 4 817 66
          DQ336698.1 - 4 817 66
          EU048339.1 - 1 210 66
          EU668336.1 - 4 829 66
          FM205711.1 - 4 829 66
          HE663398.1 - 5 778 66
          HG327115.1 - 4 829 66
          HG327117.1 - 4 829 66
          HG327119.1 - 4 829 66
          HG327120.1 - 4 829 66
          HG327121.1 - 4 829 66
          HG514457.1 - 4 829 66
          L04695.1 - 4 829 66
          L04696.1 - 4 829 66
          L07743.1 - 4 829 66
          L07950.1 - 4 829 66
          L09735.1 - 4 829 66
          L09736.1 - 4 829 66
          L11570.1 - 4 829 66
          L11571.1 - 4 829 66
          L11603.1 - 4 829 66
          L11604.1 - 4 829 66
          L11666.1 - 4 829 66
          L15005.1 - 4 829 66
          L17005.1 - 4 829 66
          L19923.1 - 4 829 66
          L19937.1 - 4 829 66
          L20088.1 - 4 824 66
          L20089.1 - 4 824 66
          L20090.1 - 5 824 66
          L22027.1 - 4 829 66
          L22028.1 - 4 829 66
          L32862.1 - 4 829 66
          L33922.1 - 4 829 66
          L36318.1 - 4 829 66
          L36591.1 - 4 829 66
          L37880.1 - 4 829 66
          L37881.1 - 4 829 66
          L38504.1 - 4 829 66
          L41214.1 - 5 829 66
          L41925.1 - 5 829 66
          M12678.1 - 4 820 66
          M15470.1 - 4 820 66
          M24033.1 - 4 829 66
          M28203.1 - 4 818 66
          M28204.1 - 4 793 66
          M28205.1 - 5 829 66
          M32317.1 - 4 829 66
          M32318.1 - 5 829 66
          M32319.1 - 5 829 66
          M32320.1 - 4 829 66
          M62852.1 - 3 757 66
          M75138.1 - 4 829 66
          M77774.1 - 5 829 66
          M77775.1 - 4 829 66
          M77776.1 - 4 829 66
          M77777.1 - 4 829 66
          M77778.1 - 4 829 66
          M81798.1 - 4 829 66
          M83191.1 - 3 757 66
          M83192.1 - 3 757 66
          M83193.1 - 3 757 66
          M83194.1 - 3 757 66
          M83195.1 - 3 757 66
          M84380.1 - 4 829 66
          M84381.1 - 4 829 66
          M84382.1 - 4 829 66
          M84383.1 - 4 829 66
          M84384.1 - 4 829 66
          M84385.1 - 4 829 66
          M84694.1 - 4 829 66
          M86403.1 - 4 805 66
          U01848.1 - 4 829 66
          U03698.1 - 4 829 66
          U03859.1 - 4 829 66
          U04243.1 - 4 829 66
          U04244.1 - 4 829 66
          U04245.1 - 4 829 66
          U06697.1 - 3 756 66
          U06862.1 - 4 829 66
          U09864.1 - 4 829 66
          U09912.1 - 4 829 66
          U11261.1 - 4 829 66
          U11262.1 - 4 829 66
          U11263.1 - 4 829 66
          U11264.1 - 4 829 66
          U11265.1 - 4 829 66
          U11266.1 - 4 829 66
          U11267.1 - 4 829 66
          U11268.1 - 4 829 66
          U11269.1 - 4 829 66
          U14943.1 - 4 829 66
          U17572.1 - 4 829 66
          U21052.1 - 4 829 66
          U21053.1 - 4 829 66
          U29057.1 - 4 829 66
          U29083.1 - 4 829 66
          U29480.1 - 4 829 66
          U30904.1 - 4 829 66
          U30936.1 - 4 829 66
          U31971.1 - 4 829 66
          U35431.1 - 4 829 66
          U35734.1 - 4 829 66
          U36392.1 - 4 829 66
          U36492.1 - 4 829 66
          U38800.1 - 4 829 66
          U40498.1 - 4 829 66
          U50710.1 - 4 829 66
          U57966.1 - 4 829 66
          U63653.1 - 4 829 66
          U64801.1 - 4 829 66
          U70528.1 - 4 829 66
          U70529.1 - 4 829 66
          U75533.1 - 4 829 66
          U80945.1 - 4 829 66
          X61706.1 - 4 829 66
          X61708.1 - 5 829 66
          X61709.1 - 4 829 66
          X61710.1 - 4 829 66
          X64366.1 - 4 829 66
          X64454.1 - 4 829 66
          X73578.1 - 4 829 66
          X75953.1 - 5 829 66
          X81363.1 - 4 829 66
          X84725.1 - 4 829 66
          X90390.1 - 4 829 66
          X90391.1 - 4 829 66
          X91749.1 - 4 829 66
          Y08994.1 - 5 829 66
          Y08995.1 - 4 829 66
          Y09058.1 - 4 829 66
          Y09118.1 - 4 829 66
          Z15143.1 - 5 829 66
          Z22651.1 - 4 829 66
          Z33453.1 - 4 830 66
          Gene Symbol: BBX
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY527413.1 - 4 829 66
          Gene Symbol: BCKDHA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA J04474.1 - 1 1794 66
          Gene Symbol: CCT6A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AM295155.1 - 1 1810 66
          Gene Symbol: CELF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF267534.1 - 956 66
          Gene Symbol: CEP41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF429309.1 - 1 2053 66
          Gene Symbol: CPSF6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA X67337.1 - 1 1844 66
          Gene Symbol: FOSB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA L49169.1 - 1 1976 66
          Gene Symbol: FUT8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA Y17976.1 - 1 2062 66
          Gene Symbol: GCOM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY334560.1 - 2 1811 66
          Gene Symbol: IFI16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY138863.1 - 1 1814 66
          Gene Symbol: LTF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY165046.1 - 1 2038 66
          Gene Symbol: NOTCH3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA PQ115184.1 - 1 2399 66
          Gene Symbol: PLEK
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA X07743.1 - 1 2018 66
          Gene Symbol: PTGIS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF297051.1 - 1571 66
          Gene Symbol: SBDS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY169963.1 - 1 1777 66
          Gene Symbol: TCF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FR748218.1 - 1129 66
          Gene Symbol: TOP3B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF017146.1 - 2 2117 66
          Gene Symbol: TRIM22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA HQ842635.1 - 1129 66
          Gene Symbol: TYK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA MN842724.1 - 1 2061 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HLA-B
          Click here to find siRNAs for Gene BBX
          Click here to find siRNAs for Gene BCKDHA
          Click here to find siRNAs for Gene CCT6A
          Click here to find siRNAs for Gene CELF1
          Click here to find siRNAs for Gene CEP41
          Click here to find siRNAs for Gene CPSF6
          Click here to find siRNAs for Gene FOSB
          Click here to find siRNAs for Gene FUT8
          Click here to find siRNAs for Gene GCOM1
          Click here to find siRNAs for Gene IFI16
          Click here to find siRNAs for Gene LTF
          Click here to find siRNAs for Gene NOTCH3
          Click here to find siRNAs for Gene PLEK
          Click here to find siRNAs for Gene PTGIS
          Click here to find siRNAs for Gene SBDS
          Click here to find siRNAs for Gene TCF4
          Click here to find siRNAs for Gene TOP3B
          Click here to find siRNAs for Gene TRIM22
          Click here to find siRNAs for Gene TYK2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          adaptive immune response
          immune system process
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class Ib
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-independent
          regulation of T cell anergy
          defense response
          immune response
          detection of bacterium
          antigen processing and presentation
          regulation of interleukin-12 production
          regulation of interleukin-6 production
          protection from natural killer cell mediated cytotoxicity
          innate immune response
          regulation of dendritic cell differentiation
          receptor binding
          protein binding
          peptide antigen binding
          TAP binding
          chaperone binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0