Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00431846_m1
          See other AQP8 GE Assays ›
          Gene Symbol:
          Aqp8
          Gene Name:
          aquaporin 8
          Gene Aliases:
          AI255744, AQP-8
          Chromosome Location:
          Chr.7: 123462294 - 123468003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00431846_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aqp8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109045.1 NP_001102515.1 4 - 5 754 74
          NM_007474.2 NP_031500.1 4 - 5 757 74
          GenBank mRNA AF018952.1 - 4 - 5 739 74
          AK146660.1 - 3 - 4 436 74
          BC010982.1 - 4 - 5 699 4224850 74
          CF617211.1 - 4 - 5 660 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001774.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aqp8

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          water transport
          cellular water homeostasis
          canalicular bile acid transport
          glycerol transport
          carbohydrate transmembrane transport
          ion transmembrane transport
          transmembrane transport
          transporter activity
          water channel activity
          glycerol channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078955 BC010982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078955 AF018952.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078955 BY713653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078955 CF617211.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078955 NM_001109045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2318584 AK146660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2318584 BC010982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2318584 NM_001109045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2318584 NM_007474.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855565 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162344 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078955 BC010982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078955 AF018952.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078955 BY713653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078955 CF617211.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078955 NM_001109045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2318584 AK146660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2318584 BC010982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2318584 NM_001109045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2318584 NM_007474.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P183261 AK146660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P183261 BC010982.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P183261 AF018952.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P183261 NM_001109045.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P183261 NM_007474.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25484542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162344 - -
          Whole Mouse Genome G4122A A_51_P183261 AK146660.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183261 BC010982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183261 AF018952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183261 NM_001109045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183261 NM_007474.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116602 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06179219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06179215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06179216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25484595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06024938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06274553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05932495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06547248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06274549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06123254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25484550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557306 - -
          Affymetrix HT MG-430 PM Array Plate 1417828_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557300 - -
          Mouse Genome 430 2.0 Array 1417828_PM_at - -
          Mouse Genome 430 2.0 Array 1417828_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11833_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11833_at - -
          Affymetrix Mouse Clariom S Array PSR0700015174.mm.1 TC0700001774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015174.mm.1 NM_001109045.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015174.mm.1 NM_001109045.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015174.mm.1 NM_007474.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015174.mm.1 NM_007474.2 Refseq
          Mouse Expression Set 430 1417828_PM_at - -
          Mouse Expression Set 430 1417828_at - -
          Murine Genome U74v2 Set 102200_PM_at - -
          Murine Genome U74v2 Set 102200_at - -
          Mouse Genome 430A 2.0 Array 1417828_PM_at - -
          Mouse Genome 430A 2.0 Array 1417828_at - -
          Affymetrix HT Mouse Genome 430A Array 1417828_at - -
          Affymetrix HT Mouse Genome 430A Array 1417828_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 467024 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756379 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756379 - -
          mouse-6 v1.1 (Illumina) scl32080.5_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32080.5_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2756379 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2854157 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2756379 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854157 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline