Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433752_m1
          See other GRIA1 GE Assays ›
          Gene Symbol:
          Gria1
          Gene Name:
          glutamate receptor, ionotropic, AMPA1 (alpha 1)
          Gene Aliases:
          2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1
          Chromosome Location:
          Chr.11: 57011571 - 57330244 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433752_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gria1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113325.2 NP_001106796.1 4 - 5 1057 66
          NM_001252403.1 NP_001239332.1 4 - 5 684 66
          NM_008165.4 NP_032191.2 4 - 5 1057 66
          GenBank mRNA AB102773.1 - 4 - 5 641 66
          AB102774.1 - 4 - 5 641 66
          AK049958.1 - 4 - 5 867 66
          AK135107.1 - 4 - 5 685 66
          BC056397.1 - 4 - 5 719 6842391 66
          BC060702.1 - 4 - 5 640 66
          JN180918.1 - 708 66
          JN180919.1 - 4 - 5 639 66
          JN180920.1 - 4 - 5 529 66
          X57497.1 - 4 - 5 904 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000654.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gria1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor recycling
          transport
          ion transport
          chemical synaptic transmission
          long-term memory
          response to toxic substance
          neuronal action potential
          receptor internalization
          ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          positive regulation of membrane potential
          regulation of synaptic plasticity
          modulation of synaptic transmission
          positive regulation of synaptic transmission
          long term synaptic depression
          G-protein alpha-subunit binding
          receptor activity
          ionotropic glutamate receptor activity
          AMPA glutamate receptor activity
          ion channel activity
          extracellular-glutamate-gated ion channel activity
          protein binding
          adenylate cyclase binding
          protein kinase binding
          protein domain specific binding
          PDZ domain binding
          small GTPase binding
          myosin V binding
          G-protein beta-subunit binding
          beta-2 adrenergic receptor binding
          identical protein binding
          protein kinase A binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_001113325.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_001113325.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_001252403.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_001252403.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_008165.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 NM_008165.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005286.mm.1 TC1100000654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_001113325.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_001113325.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_001252403.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_001252403.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_008165.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 NM_008165.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005285.mm.1 TC1100000654.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline