Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435104_m1
          See other MYBPC3 GE Assays ›
          Gene Symbol:
          Mybpc3
          Gene Name:
          myosin binding protein C, cardiac
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 91118114 - 91136518 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435104_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mybpc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008653.2 NP_032679.2 20 - 21 1962 75
          GenBank mRNA AF097333.1 - 20 - 21 1913 75
          AK146661.1 - 20 - 21 1963 75
          AK146886.1 - 20 - 21 1963 75
          AK169302.1 - 19 - 20 1940 75
          AK169306.1 - 19 - 20 1940 75
          BC094945.1 - 20 - 21 1959 75
          BC131672.1 - 20 - 21 1938 75
          BC154408.1 - 20 - 21 1983 40110890 75

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mybpc3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          heart morphogenesis
          muscle contraction
          cell adhesion
          regulation of heart contraction
          myosin filament assembly
          sarcomere organization
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          actin binding
          structural constituent of cytoskeleton
          structural constituent of muscle
          myosin binding
          myosin heavy chain binding
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011862 AK146661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011862 AK146886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011862 AK169302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011862 AK169306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011862 BC094945.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175570 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164457 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150662 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011862 AK146661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011862 AK146886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011862 AK169302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011862 AK169306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011862 BC094945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420789 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04603037 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04603087 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164457 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04603023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150662 - -
          Whole Mouse Genome G4122A A_51_P465449 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102341 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164457 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150662 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367257 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01367232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01533242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01875670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01757377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01875673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01395104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01341972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473833 - -
          Affymetrix HT MG-430 PM Array Plate 1418551_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473814 - -
          Mouse Genome 430 2.0 Array 1418551_at - -
          Mouse Genome 430 2.0 Array 1418551_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17868_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17868_at - -
          Mouse Expression Set 430 1418551_at - -
          Mouse Expression Set 430 1418551_PM_at - -
          Murine Genome U74v2 Set 160582_PM_at - -
          Murine Genome U74v2 Set 160582_at - -
          Mouse Genome 430A 2.0 Array 1418551_PM_at - -
          Mouse Genome 430A 2.0 Array 1418551_at - -
          Affymetrix HT Mouse Genome 430A Array 1418551_at - -
          Affymetrix HT Mouse Genome 430A Array 1418551_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 635695 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 840048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689136 - -
          mouse-6 v1.1 (Illumina) scl017868.31_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 840048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017868.31_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689136 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline