Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008861.3 | NP_032887.3 | 1 - 2 | 770 | 84 | |
GenBank mRNA | AF014010.1 | - | 1 - 2 | 673 | 84 | |
AK053502.1 | - | 1 - 2 | 675 | 84 | ||
AK128961.1 | - | 2 - 3 | 544 | 84 | ||
AK164673.1 | - | 1 - 2 | 661 | 84 | ||
BC053058.1 | - | 1 - 2 | 777 | 84 | ||
BC062969.1 | - | 1 - 2 | 777 | 84 | ||
FJ609779.1 | - | 1 - 2 | 686 | 84 | ||
Y13278.1 | - | 1 - 2 | 686 | 84 | ||
Y14120.1 | - | 1 - 2 | 436 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |