Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437157_g1
          See other TNNI2 GE Assays ›
          Gene Symbol:
          Tnni2
          Gene Name:
          troponin I, skeletal, fast 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 142440494 - 142444410 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437157_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnni2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009405.2 NP_033431.1 4 - 5 210 62
          GenBank mRNA BC028515.1 - 5 - 6 248 1448494 62
          J04992.1 - 4 - 5 210 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002076.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnni2

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          regulation of muscle contraction
          positive regulation of transcription, DNA-templated
          actin binding
          troponin T binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040893 NM_009405.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040893 BC028515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040893 J04992.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568922 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166634 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040893 NM_009405.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040893 BC028515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040893 J04992.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355122 NM_009405.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355122 BC028515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355122 J04992.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25594685 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166634 - -
          Whole Mouse Genome G4122A A_51_P355122 NM_009405.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355122 BC028515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355122 J04992.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112279 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458739 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06296911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458742 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06458752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06296928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25594618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06296936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06296928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06239562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06051294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06051299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06051291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06051289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06526703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25594774 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559205 - -
          Affymetrix HT MG-430 PM Array Plate 1416889_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438609_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1438608_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559200 - -
          Mouse Genome 430 2.0 Array 1438609_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416889_at - -
          Mouse Genome 430 2.0 Array 1438608_PM_at - -
          Mouse Genome 430 2.0 Array 1438608_at - -
          Mouse Genome 430 2.0 Array 1438609_x_at - -
          Mouse Genome 430 2.0 Array 1416889_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21953_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21953_at - -
          Affymetrix Mouse Clariom S Array PSR0700018655.mm.1 TC0700002076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018655.mm.1 NM_009405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018655.mm.1 NM_009405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018647.mm.1 TC0700002076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018647.mm.1 NM_009405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018647.mm.1 NM_009405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018658.mm.1 TC0700002076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018658.mm.1 NM_009405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018658.mm.1 NM_009405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018660.mm.1 TC0700002076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018660.mm.1 NM_009405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018660.mm.1 NM_009405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018654.mm.1 TC0700002076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018654.mm.1 NM_009405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018654.mm.1 NM_009405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018643.mm.1 TC0700002076.mm.1 MTA TC
          Mouse Expression Set 430 1438608_at - -
          Mouse Expression Set 430 1438608_PM_at - -
          Mouse Expression Set 430 1438609_x_at - -
          Mouse Expression Set 430 1416889_at - -
          Mouse Expression Set 430 1438609_PM_x_at - -
          Mouse Expression Set 430 1416889_PM_at - -
          Murine Genome U74v2 Set 93532_at - -
          Murine Genome U74v2 Set 93532_PM_at - -
          Mouse Genome 430A 2.0 Array 1416889_at - -
          Mouse Genome 430A 2.0 Array 1438609_x_at - -
          Mouse Genome 430A 2.0 Array 1416889_PM_at - -
          Mouse Genome 430A 2.0 Array 1438608_PM_at - -
          Mouse Genome 430A 2.0 Array 1438609_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438608_at - -
          Affymetrix HT Mouse Genome 430A Array 1438608_at - -
          Affymetrix HT Mouse Genome 430A Array 1416889_at - -
          Affymetrix HT Mouse Genome 430A Array 1438608_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438609_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416889_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438609_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 796046 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910739 - -
          Illumina mouse-6 v1.0 expression beadchip 5910739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2481133 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2481133 - -
          mouse-6 v1.1 (Illumina) scl31869.6.1_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31869.6.1_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2481133 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2481133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline