Search
Search

Bmp15| Gene Symbol: Bmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009757.4 | NP_033887.1 | 1 - 2 | 689 | 64 | |
| GenBank mRNA | AF082348.1 | - | 1 - 2 | 572 | 64 | |
| AJ132406.1 | - | 1 - 2 | 329 | 64 | ||
| AK028735.1 | - | 1 - 2 | 375 | 64 | ||
| AK135757.1 | - | 1 - 2 | 596 | 64 | ||
| AK135882.1 | - | 1 - 2 | 692 | 64 | ||
| AK136127.1 | - | 1 - 2 | 83 | 64 | ||
| AK136129.1 | - | 1 - 2 | 83 | 64 | ||
| AK139773.1 | - | 1 - 2 | 690 | 64 | ||
| AK139847.1 | - | 1 - 2 | 691 | 64 | ||
| AK139936.1 | - | 1 - 2 | 645 | 64 | ||
| BC055363.1 | - | 1 - 2 | 686 | 30051425 | 64 | |
Set Membership: |
|