Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438567_s1
          See other ADAM24 GE Assays ›
          Gene Symbol:
          Adam24
          Gene Name:
          a disintegrin and metallopeptidase domain 24 (testase 1)
          Gene Aliases:
          Dtgn5
          Chromosome Location:
          Chr.8: 40675094 - 40682199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438567_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010086.4 NP_034216.3 2 373 92
          GenBank mRNA AF167402.1 - 1 361 92
          AK029528.1 - 2 374 92
          AK133199.1 - 2 390 92
          BC145785.1 - 1 262 40131088 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000477.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam24

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          multicellular organism development
          spermatogenesis
          cell differentiation
          prevention of polyspermy
          metalloendopeptidase activity
          integrin binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 BC145785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AK029528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AK133199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 NM_010086.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 BC145785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AK029528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AK133199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 NM_010086.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159325 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109558 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 BC145785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AK029528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AK133199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 NM_010086.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AF167402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 BC145785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AK029528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AK133199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 NM_010086.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 BC145785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AK029528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AK133199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 NM_010086.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 BC145785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AK029528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AK133199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 NM_010086.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25807162 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109558 - -
          Whole Mouse Genome G4122A A_51_P379083 BC145785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AK029528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AK133199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 NM_010086.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AF167402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 BC145785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AK029528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AK133199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 NM_010086.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379083 AF167402.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109910 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07100073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07421979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07100064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07100068 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07100065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25807158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06846593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07405985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06871587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07006635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06678360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06933207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07447365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25807193 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10571435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571433 - -
          Affymetrix HT MG-430 PM Array Plate 1449513_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571432 - -
          Mouse Genome 430 2.0 Array 1449513_PM_at - -
          Mouse Genome 430 2.0 Array 1449513_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13526_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13526_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800003306.mm.1 NM_010086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003306.mm.1 TC0800000477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800003305.mm.1 NM_010086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003305.mm.1 TC0800000477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800003304.mm.1 NM_010086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003304.mm.1 TC0800000477.mm.1 MTA TC
          Mouse Expression Set 430 1449513_at - -
          Mouse Expression Set 430 1449513_PM_at - -
          Murine Genome U74v2 Set 166149_PM_at - -
          Murine Genome U74v2 Set 166149_at - -
          Mouse Genome 430A 2.0 Array 1449513_PM_at - -
          Mouse Genome 430A 2.0 Array 1449513_at - -
          Affymetrix HT Mouse Genome 430A Array 1449513_at - -
          Affymetrix HT Mouse Genome 430A Array 1449513_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416281 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670390 - -
          Illumina mouse-6 v1.0 expression beadchip 670390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218770 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218770 - -
          mouse-6 v1.1 (Illumina) scl33877.2_528-S - -
          Illumina Sentrix mouse-6.v1 bead array 670390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33877.2_528-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218770 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline