Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439631_m1
          See other IL3 GE Assays ›
          Gene Symbol:
          Il3
          Gene Name:
          interleukin 3
          Gene Aliases:
          BPA, Csfmu, HCGF, Il-3, MCGF, PSF
          Chromosome Location:
          Chr.11: 54265085 - 54267279 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010556.4 NP_034686.2 2 - 3 238 85
          GenBank mRNA A02046.1 - 2 - 3 236 85
          AK153634.1 - 2 - 3 239 85
          BC125552.1 - 2 - 3 233 40130067 85
          BC125554.1 - 2 - 3 233 40130069 85
          K01668.1 - 2 - 3 236 85
          K01850.1 - 2 - 3 236 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002759.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          response to hypoxia
          positive regulation of protein phosphorylation
          positive regulation of myeloid leukocyte differentiation
          regulation of glycolytic process
          immune response
          positive regulation of cell proliferation
          response to hormone
          regulation of gene expression
          negative regulation of autophagy
          cytokine-mediated signaling pathway
          hemopoiesis
          negative regulation of mast cell apoptotic process
          myeloid cell apoptotic process
          regulation of protein dephosphorylation
          negative regulation of apoptotic process
          positive regulation of JNK cascade
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of mast cell proliferation
          extrinsic apoptotic signaling pathway in absence of ligand
          cytokine activity
          interleukin-3 receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985985 NM_010556.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985985 AK153634.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144477 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161645 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985985 NM_010556.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985985 AK153634.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98630 A02046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98630 NM_010556.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98630 AK153634.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98630 BC125552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98630 BC125554.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27809148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161645 - -
          Whole Mouse Genome G4122A A_52_P98630 A02046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98630 NM_010556.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98630 AK153634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98630 BC125552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98630 BC125554.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129121 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161645 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385199 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27809080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09108689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09108700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09108720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09370393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09569525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09108717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27809179 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385922 - -
          Affymetrix HT MG-430 PM Array Plate 1450566_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385918 - -
          Mouse Genome 430 2.0 Array 1450566_PM_at - -
          Mouse Genome 430 2.0 Array 1450566_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16187_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16187_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100025018.mm.1 NM_010556.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025018.mm.1 NM_010556.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025018.mm.1 TC1100002759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025017.mm.1 NM_010556.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025017.mm.1 NM_010556.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025017.mm.1 TC1100002759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025019.mm.1 NM_010556.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025019.mm.1 NM_010556.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025019.mm.1 TC1100002759.mm.1 MTA TC
          Mouse Expression Set 430 1450566_at - -
          Mouse Expression Set 430 1450566_PM_at - -
          Murine Genome U74v2 Set 94086_at - -
          Murine Genome U74v2 Set 94086_PM_at - -
          Mouse Genome 430A 2.0 Array 1450566_PM_at - -
          Mouse Genome 430A 2.0 Array 1450566_at - -
          Affymetrix HT Mouse Genome 430A Array 1450566_at - -
          Affymetrix HT Mouse Genome 430A Array 1450566_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 391914 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2480280 - -
          Illumina mouse-6 v1.0 expression beadchip 2480280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247309 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247309 - -
          mouse-6 v1.1 (Illumina) scl40215.5.1_93-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40215.5.1_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247309 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247309 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline