Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00443098_g1
          See other PTGER1 GE Assays ›
          Gene Symbol:
          Ptger1
          Gene Name:
          prostaglandin E receptor 1 (subtype EP1)
          Gene Aliases:
          42kDa, EP1, Ptgerep1
          Chromosome Location:
          Chr.8: 83666530 - 83670103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00443098_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptger1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013641.2 NP_038669.1 2 - 3 1247 85
          GenBank mRNA AK053616.1 - 86
          BC138739.1 - 2 - 3 1209 8861761 85
          BC138744.1 - 2 - 3 1209 8861766 85
          D16338.1 - 2 - 3 1024 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000931.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptger1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          response to lipopolysaccharide
          signal transducer activity
          G-protein coupled receptor activity
          prostaglandin receptor activity
          prostaglandin E receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015929 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015930 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015925 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995473 CX568360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995473 NM_177262.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995473 NM_001199593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995473 AK218198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995473 AK164746.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629137 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171854 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995473 CX568360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995473 NM_177262.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995473 NM_001199593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995473 AK218198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995473 AK164746.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26015911 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171854 - -
          Whole Mouse Genome G4122A A_51_P144303 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07243836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015929 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26015930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26015907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07173179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07433926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07243847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07243836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07433933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07433878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26015939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573213 - -
          Affymetrix HT MG-430 PM Array Plate 1450491_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445445_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10573210 - -
          Mouse Genome 430 2.0 Array 1436542_PM_at - -
          Mouse Genome 430 2.0 Array 1450491_PM_at - -
          Mouse Genome 430 2.0 Array 1436851_PM_at - -
          Mouse Genome 430 2.0 Array 1445445_PM_s_at - -
          Mouse Genome 430 2.0 Array 1436542_at - -
          Mouse Genome 430 2.0 Array 1458674_PM_at - -
          Mouse Genome 430 2.0 Array 1436851_at - -
          Mouse Genome 430 2.0 Array 1450491_at - -
          Mouse Genome 430 2.0 Array 1458674_at - -
          Mouse Genome 430 2.0 Array 1445445_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19216_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19216_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800006880.mm.1 NM_013641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800006880.mm.1 NM_013641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800006880.mm.1 TC0800000931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800006882.mm.1 NM_013641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800006882.mm.1 NM_013641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800006882.mm.1 TC0800000931.mm.1 MTA TC
          Mouse Expression Set 430 1436851_at - -
          Mouse Expression Set 430 1436851_PM_at - -
          Mouse Expression Set 430 1450491_PM_at - -
          Mouse Expression Set 430 1450491_at - -
          Mouse Expression Set 430 1445445_s_at - -
          Mouse Expression Set 430 1445445_PM_s_at - -
          Murine Genome U74v2 Set 99998_at - -
          Murine Genome U74v2 Set 99997_at - -
          Murine Genome U74v2 Set 99998_PM_at - -
          Murine Genome U74v2 Set 99997_PM_at - -
          Murine Genome U74v2 Set 166579_i_at - -
          Murine Genome U74v2 Set 166579_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1436851_PM_at - -
          Mouse Genome 430A 2.0 Array 1436851_at - -
          Mouse Genome 430A 2.0 Array 1450491_at - -
          Mouse Genome 430A 2.0 Array 1450491_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450491_at - -
          Affymetrix HT Mouse Genome 430A Array 1450491_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445445_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1445445_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485989 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100132 - -
          Illumina mouse-6 v1.0 expression beadchip 101990603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216138 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2432308 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216138 - -
          mouse-6 v1.1 (Illumina) scl0019216.2_175-S - -
          mouse-6 v1.1 (Illumina) scl33603.4_508-S - -
          Illumina Sentrix mouse-6.v1 bead array 101990603 - -
          Illumina Sentrix mouse-6.v1 bead array 6100132 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0019216.2_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101990603 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline