Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445039_m1
          See other CCL28 GE Assays ›
          Gene Symbol:
          Ccl28
          Gene Name:
          chemokine (C-C motif) ligand 28
          Gene Aliases:
          CCK1, MEC, Scya28
          Chromosome Location:
          Chr.13: 119610551 - 119654359 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445039_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020279.3 NP_064675.1 2 - 3 242 89
          GenBank mRNA AF220238.1 - 2 - 3 188 89
          BC040510.1 - 2 - 3 254 89
          BC055864.1 - 2 - 3 245 4240191 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001350.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Chemokine Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl28

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-matrix adhesion
          chemotaxis
          immune response
          positive regulation of cytosolic calcium ion concentration
          cell chemotaxis
          negative regulation of leukocyte tethering or rolling
          cytokine activity
          chemokine activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175915 BC040510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175915 NM_020279.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175915 BC040510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175915 NM_020279.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276560 BC040510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276560 BC055864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276560 NM_020279.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276560 AF220238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276560 BC040510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276560 BC055864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276560 NM_020279.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276560 AF220238.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118803 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598204 - -
          Affymetrix HT MG-430 PM Array Plate 1450218_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455577_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450217_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598203 - -
          Mouse Genome 430 2.0 Array 1455577_at - -
          Mouse Genome 430 2.0 Array 1450218_at - -
          Mouse Genome 430 2.0 Array 1450217_at - -
          Mouse Genome 430 2.0 Array 1450218_PM_at - -
          Mouse Genome 430 2.0 Array 1455577_PM_at - -
          Mouse Genome 430 2.0 Array 1450217_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300008482.mm.1 NM_020279.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008482.mm.1 NM_020279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008482.mm.1 TC1300001350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008483.mm.1 NM_020279.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008483.mm.1 NM_020279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008483.mm.1 TC1300001350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008484.mm.1 NM_020279.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008484.mm.1 NM_020279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008484.mm.1 TC1300001350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008485.mm.1 NM_020279.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008485.mm.1 NM_020279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008485.mm.1 TC1300001350.mm.1 MTA TC
          Mouse Expression Set 430 1450218_PM_at - -
          Mouse Expression Set 430 1450217_at - -
          Mouse Expression Set 430 1450217_PM_at - -
          Mouse Expression Set 430 1450218_at - -
          Mouse Expression Set 430 1455577_PM_at - -
          Mouse Expression Set 430 1455577_at - -
          Mouse Genome 430A 2.0 Array 1450217_at - -
          Mouse Genome 430A 2.0 Array 1450217_PM_at - -
          Mouse Genome 430A 2.0 Array 1450218_at - -
          Mouse Genome 430A 2.0 Array 1450218_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450217_at - -
          Affymetrix HT Mouse Genome 430A Array 1450218_at - -
          Affymetrix HT Mouse Genome 430A Array 1450217_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450218_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455577_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455577_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690593 - -
          Illumina mouse-6 v1.0 expression beadchip 2690593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218590 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218590 - -
          mouse-6 v1.1 (Illumina) scl0056838.1_132-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056838.1_132-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218590 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline