Search
Search
| Gene Symbol: Tgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031199.3 | NP_112476.1 | 2 - 3 | 354 | 111 | |
| GenBank mRNA | AK010785.1 | - | 2 - 3 | 382 | 111 | |
| AK162680.1 | - | 2 - 3 | 119 | 111 | ||
| AK166321.1 | - | 2 - 3 | 287 | 111 | ||
| BC132185.1 | - | 2 - 3 | 152 | 40130462 | 111 | |
| BC132211.1 | - | 2 - 3 | 152 | 40130488 | 111 | |
| M92420.1 | - | 2 - 4 | 104 | 111 | ||
| U65016.1 | - | 2 - 3 | 189 | 111 | ||
Set Membership: |
|