Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00450896_m1
          See other PQBP1 GE Assays ›
          Gene Symbol:
          Pqbp1
          Gene Name:
          polyglutamine binding protein 1
          Gene Aliases:
          PQBP-1, Sfc2, npw38
          Chromosome Location:
          Chr.X: 7894519 - 7899269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00450896_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pqbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252528.1 NP_001239457.1 2 - 3 122 86
          NM_001252529.1 NP_001239458.1 2 - 3 188 86
          NM_019478.4 NP_062351.2 2 - 3 317 86
          GenBank mRNA AJ250406.1 - 1 - 2 68 86
          AK077652.1 - 2 - 3 147 86
          BC009657.1 - 1 - 2 90 3484619 86
          BC051673.1 - 2 - 3 127 30015428 86
          BC157976.1 - 2 - 3 222 9088137 86
          BQ550428.1 - 2 - 3 193 86
          CJ143105.1 - 2 - 3 123 86
          CN718083.1 - 1 - 2 102 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001784.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pqbp1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          alternative mRNA splicing, via spliceosome
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          RNA splicing
          neuron projection development
          stress granule assembly
          regulation of RNA splicing
          regulation of dendrite morphogenesis
          cilium morphogenesis
          protein C-terminus binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32240780 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070250 BC157976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 AK077652.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 BC009657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 BC051673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 NM_019478.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 AK077652.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC009657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC051673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC157976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 NM_019478.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717461 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101557 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 AK077652.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 BC009657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 BC051673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 NM_019478.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 AK077652.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC009657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC051673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC157976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 NM_019478.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135322 AJ250406.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240771 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101557 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09227085 - -
          Whole Mouse Genome G4122A A_51_P135322 AJ250406.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112808 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101557 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14941590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14941581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32240780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14517909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14772639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14517915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14772636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14517913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14534934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14941581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603381 - -
          Affymetrix HT MG-430 PM Array Plate 1417731_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10603373 - -
          Mouse Genome 430 2.0 Array 1456086_x_at - -
          Mouse Genome 430 2.0 Array 1417731_PM_at - -
          Mouse Genome 430 2.0 Array 1417731_at - -
          Mouse Genome 430 2.0 Array 1456086_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54633_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54633_at - -
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_001252528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_001252528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_001252529.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_001252529.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_019478.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 NM_019478.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011300.mm.1 TC0X00001784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011307.mm.1 NM_019478.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011307.mm.1 NM_019478.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011307.mm.1 TC0X00001784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011305.mm.1 NM_019478.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011305.mm.1 NM_019478.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011305.mm.1 TC0X00001784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_001252528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_001252528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_001252529.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_001252529.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_019478.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 NM_019478.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011299.mm.1 TC0X00001784.mm.1 MTA TC
          Mouse Expression Set 430 1456086_PM_x_at - -
          Mouse Expression Set 430 1456086_x_at - -
          Mouse Expression Set 430 1417731_PM_at - -
          Mouse Expression Set 430 1417731_at - -
          Murine Genome U74v2 Set 93010_at - -
          Murine Genome U74v2 Set 93010_PM_at - -
          Mouse Genome 430A 2.0 Array 1417731_at - -
          Mouse Genome 430A 2.0 Array 1456086_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1417731_PM_at - -
          Mouse Genome 430A 2.0 Array 1456086_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1417731_at - -
          Affymetrix HT Mouse Genome 430A Array 1417731_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910025 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520603 - -
          Illumina mouse-6 v1.0 expression beadchip 6520603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520603 - -
          mouse-6 v1.1 (Illumina) scl54442.9.1_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54442.9.1_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2883110 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2883110 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677391 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54442.9.1_77-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline