Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451486_s1
          See other OLFR480 GE Assays ›
          Gene Symbol:
          Olfr480
          Gene Name:
          olfactory receptor 480
          Gene Aliases:
          MOR204-32, Ors25, S25
          Chromosome Location:
          Chr.7: 108065768 - 108066796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451486_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr480
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020291.1 NP_064687.1 1 943 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr480

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032089 NM_020291.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116116 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032089 NM_020291.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130567 NM_020291.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402156 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116116 - -
          Whole Mouse Genome G4122A A_51_P130567 NM_020291.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133742 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402146 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566673 - -
          Affymetrix HT MG-430 PM Array Plate 1422351_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566672 - -
          Mouse Genome 430 2.0 Array 1422351_at - -
          Mouse Genome 430 2.0 Array 1422351_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56861_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56861_PM_at - -
          Mouse Expression Set 430 1422351_at - -
          Mouse Expression Set 430 1422351_PM_at - -
          Mouse Genome 430A 2.0 Array 1422351_at - -
          Mouse Genome 430A 2.0 Array 1422351_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422351_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422351_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 695268 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980685 - -
          Illumina mouse-6 v1.0 expression beadchip 6980685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734135 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734135 - -
          mouse-6 v1.1 (Illumina) scl8834.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8834.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2734135 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2734135 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline