Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459629_g1
          See other TNFRSF25 GE Assays ›
          Gene Symbol:
          Tnfrsf25
          Gene Name:
          tumor necrosis factor receptor superfamily, member 25
          Gene Aliases:
          APO-3, DDR3, DR3, LARD, TR3, TRAMP, Tnfrsf12, WSL-1, WSL-LR, Wsl
          Chromosome Location:
          Chr.4: 152115528 - 152120119 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459629_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291010.1 NP_001277939.1 9 - 10 1049 60
          NM_033042.4 NP_149031.2 6 - 7 999 60
          GenBank mRNA AF329969.1 - 9 - 10 1053 60
          AK040632.1 - 6 - 7 1668 60
          BC017526.1 - 6 - 7 999 4912933 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002013.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf25

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416613 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416645 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116305 BC017526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116305 NM_033042.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176648 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144506 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116305 BC017526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116305 NM_033042.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P460584 AF329969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P178561 AF329969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P178561 BC017526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P178561 NM_033042.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23416626 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144506 - -
          Whole Mouse Genome G4122A A_51_P178561 AF329969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178561 BC017526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178561 NM_033042.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460584 AF329969.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139302 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04029100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04029087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04029113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04029108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03540508 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04029089 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23416613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23416686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04052861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04052859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23416651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510676 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510669 - -
          Affymetrix HT MG-430 PM Array Plate 1422231_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510668 - -
          Mouse Genome 430 2.0 Array 1422231_a_at - -
          Mouse Genome 430 2.0 Array 1422231_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 85030_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 85030_at - -
          Affymetrix Mouse Clariom S Array PSR0400016491.mm.1 NM_033042.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400016491.mm.1 TC0400002013.mm.1 MTA TC
          Mouse Expression Set 430 1422231_a_at - -
          Mouse Expression Set 430 1422231_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422231_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422231_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422231_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422231_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687873 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710110 - -
          MouseWG-6 v2 Expression BeadChip 3850707 - -
          Illumina mouse-6 v1.0 expression beadchip 3850707 - -
          Illumina mouse-6 v1.0 expression beadchip 3710110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2510383 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248617 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248617 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2510383 - -
          mouse-6 v1.1 (Illumina) scl0002622.1_51-S - -
          mouse-6 v1.1 (Illumina) scl24658.9.1_90-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850707 - -
          Illumina Sentrix mouse-6.v1 bead array 3710110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24658.9.1_90-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002622.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710110 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2510383 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248617 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2510383 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248617 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24658.9.1_90-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline