Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463365_m1
          See other OPALIN GE Assays ›
          Gene Symbol:
          Opalin
          Gene Name:
          oligodendrocytic myelin paranodal and inner loop protein
          Gene Aliases:
          Tmem10, Tmp10
          Chromosome Location:
          Chr.19: 41063420 - 41077113 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opalin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153520.1 NP_705740.1 3 - 4 152 74
          GenBank mRNA AK140031.1 - 153 74
          BC060980.1 - 3 - 4 111 6533846 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001503.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opalin

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 AK078150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 AK140031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 BC060980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 NM_153520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 AK078150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 AK140031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 BC060980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624415 NM_153520.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125011 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136432 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135754 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 AK078150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 AK140031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 BC060980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 NM_153520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 AK078150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 AK140031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 BC060980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624415 NM_153520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262563 AK078150.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32112839 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09172578 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136432 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135754 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03597255 - -
          Whole Mouse Genome G4122A A_51_P262563 AK078150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 AK078150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 AK140031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 BC060980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 NM_153520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 AK078150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 AK140031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 BC060980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624415 NM_153520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110216 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135754 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136432 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14339358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32112824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14319721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14488936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14475008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14378550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14303604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14448757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14488938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14167283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14285822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32112832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467530 - -
          Affymetrix HT MG-430 PM Array Plate 1435854_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447487_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467529 - -
          Mouse Genome 430 2.0 Array 1435854_PM_at - -
          Mouse Genome 430 2.0 Array 1447487_at - -
          Mouse Genome 430 2.0 Array 1435854_at - -
          Mouse Genome 430 2.0 Array 1447487_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226115_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226115_at - -
          Affymetrix Mouse Clariom S Array PSR1900013134.mm.1 NM_153520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013134.mm.1 NM_153520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013134.mm.1 TC1900001503.mm.1 MTA TC
          Mouse Expression Set 430 1447487_PM_at - -
          Mouse Expression Set 430 1435854_at - -
          Mouse Expression Set 430 1435854_PM_at - -
          Mouse Expression Set 430 1447487_at - -
          Murine Genome U74v2 Set 135230_PM_at - -
          Murine Genome U74v2 Set 166890_at - -
          Murine Genome U74v2 Set 166890_PM_at - -
          Murine Genome U74v2 Set 135230_at - -
          Affymetrix HT Mouse Genome 430B Array 1447487_at - -
          Affymetrix HT Mouse Genome 430B Array 1435854_at - -
          Affymetrix HT Mouse Genome 430B Array 1435854_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447487_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840528 - -
          Illumina mouse-6 v1.0 expression beadchip 3840528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230025 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230025 - -
          mouse-6 v1.1 (Illumina) scl0226115.1_174-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0226115.1_174-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2824889 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2824889 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0226115.1_174-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline